This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01598051_g1
See other SUPT6H GE Assays ›
Gene Symbol:
SUPT6H
Gene Name:
SPT6 homolog, histone chaperone
Gene Aliases:
SPT6, SPT6H, emb-5
Chromosome Location:
Chr.17: 28662183 - 28702684 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01598051_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SUPT6H
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001320755.1 NP_001307684.1 34 - 35 4844 132
NM_003170.4 NP_003161.2 33 - 34 4742 132
XM_017024978.1 XP_016880467.1 35 - 36 4903 132
XM_017024979.1 XP_016880468.1 33 - 34 4980 132
XM_017024980.1 XP_016880469.1 32 - 33 4674 132
GenBank mRNA AF070532.1 - 2 - 3 191 132
AL833607.1 - 34 - 35 4845 132
BC003692.1 - 5 - 6 734 132
BC003696.1 - 18 - 19 2617 132
BC073963.1 - 6 - 7 1041 132
BC136522.1 - 33 - 34 4654 9020512 132
BC136524.1 - 33 - 34 4654 9020514 132
BC150268.1 - 33 - 34 4656 9007230 132
D79984.1 - 33 - 34 4656 132

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1700007381.hg.1
This assay targets the same location as HTA transcript cluster(s) TC17000325.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SUPT6H

Panther Classification:

Molecular Function -

RNA metabolism protein nucleic acid metabolism protein general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

chromatin remodeling
transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA processing
RNA splicing
regulation of mRNA export from nucleus
viral process
positive regulation of transcription elongation from RNA polymerase II promoter
regulation of isotype switching
regulation of mRNA processing
mRNA transport
regulation of muscle cell differentiation
negative regulation of histone H3-K27 methylation
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
histone binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR17003991.hg.1 ENST00000314616.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17003991.hg.1 NM_003170.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17003991.hg.1 TC17000325.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1700152556.hg.1 ENST00000314616.10 Ensembl
Affymetrix Human Clariom D Assay PSR1700152556.hg.1 ENST00000347486.8 Ensembl
Affymetrix Human Clariom D Assay PSR1700152556.hg.1 ENST00000581510.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700152556.hg.1 ENST00000583340.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700152556.hg.1 NM_003170.4 Refseq
Affymetrix Human Clariom D Assay PSR1700152556.hg.1 ENST00000314616.10 Ensembl
Affymetrix Human Clariom D Assay PSR1700152556.hg.1 ENST00000347486.8 Ensembl
Affymetrix Human Clariom D Assay PSR1700152556.hg.1 ENST00000581510.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700152556.hg.1 ENST00000583340.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700152556.hg.1 NM_003170.4 Refseq
Affymetrix Human Clariom D Assay PSR1700152556.hg.1 TC1700007381.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0