This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01598361_g1
See other NECTIN1 GE Assays ›
Gene Symbol:
NECTIN1
Gene Name:
nectin cell adhesion molecule 1
Gene Aliases:
CD111, CLPED1, ED4, HIgR, HV1S, HVEC, OFC7, PRR, PRR1, PVRL1, PVRR, PVRR1, SK-12, nectin-1
Chromosome Location:
Chr.11: 119638098 - 119728725 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01598361_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NECTIN1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_203285.1 NP_976030.1 7 - 8 1392 91
GenBank mRNA AF110314.1 - 7 - 8 1220 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100012535.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11002372.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NECTIN1

Gene Ontology Categories:

Function(s) Process(es)

lens morphogenesis in camera-type eye
desmosome organization
iron ion transport
immune response
cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
signal transduction
axon guidance
cell recognition
single organismal cell-cell adhesion
adherens junction organization
viral entry into host cell
regulation of synapse assembly
retina development in camera-type eye
enamel mineralization
virus receptor activity
receptor binding
protein binding
coreceptor activity
carbohydrate binding
protein homodimerization activity
virion binding
protein heterodimerization activity
cell adhesion molecule binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR11028588.hg.1 ENST00000341398.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11028588.hg.1 NM_203285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11028588.hg.1 TC11002372.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000264025.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000264025.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000264025.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000264025.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000264025.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000264025.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000264025.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000264025.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000264025.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000264025.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000264025.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000264025.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000340882.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000340882.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000340882.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000340882.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000340882.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000340882.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000340882.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000341398.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000341398.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000341398.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000341398.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000341398.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000341398.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000341398.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000341398.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000341398.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000341398.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000341398.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000524429.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000524429.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000524429.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000524429.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000524510.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000524510.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000524510.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000524510.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000524510.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000531468.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000531468.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000531468.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000532197.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000532197.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 ENST00000532197.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 NM_002855.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 NM_002855.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 NM_002855.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 NM_002855.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 NM_002855.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 NM_002855.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 NM_002855.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 NM_002855.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 NM_002855.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 NM_002855.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 NM_002855.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 NM_203285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 NM_203285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 NM_203285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 NM_203285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 NM_203285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 NM_203285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 NM_203285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 NM_203285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 NM_203285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 NM_203285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 NM_203285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 NM_203285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 NM_203285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 NM_203286.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 NM_203286.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 NM_203286.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 NM_203286.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 NM_203286.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 NM_203286.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 NM_203286.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 NM_203286.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 NM_203286.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012535.hg.1 NM_203286.1 Refseq
Affymetrix Human Clariom D Assay PSR1100196704.hg.1 ENST00000341398.6 Ensembl
Affymetrix Human Clariom D Assay PSR1100196704.hg.1 NM_203285.1 Refseq
Affymetrix Human Clariom D Assay PSR1100196704.hg.1 ENST00000341398.6 Ensembl
Affymetrix Human Clariom D Assay PSR1100196704.hg.1 NM_203285.1 Refseq
Affymetrix Human Clariom D Assay PSR1100196704.hg.1 TC1100012535.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000264025.7 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000264025.7 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000264025.7 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000264025.7 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000264025.7 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000264025.7 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000264025.7 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000264025.7 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000264025.7 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000264025.7 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000264025.7 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000264025.7 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000340882.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000340882.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000340882.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000340882.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000340882.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000340882.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000340882.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000341398.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000341398.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000341398.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000341398.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000341398.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000341398.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000341398.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000341398.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000341398.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000341398.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000341398.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000524429.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000524429.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000524429.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000524429.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000524510.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000524510.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000524510.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000524510.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000524510.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000531468.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000531468.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000531468.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000532197.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000532197.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 ENST00000532197.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012535.hg.1 NM_002855.4 Refseq
Affymetrix Human Clariom D Assay TC1100012535.hg.1 NM_002855.4 Refseq
Affymetrix Human Clariom D Assay TC1100012535.hg.1 NM_002855.4 Refseq
Affymetrix Human Clariom D Assay TC1100012535.hg.1 NM_002855.4 Refseq
Affymetrix Human Clariom D Assay TC1100012535.hg.1 NM_002855.4 Refseq
Affymetrix Human Clariom D Assay TC1100012535.hg.1 NM_002855.4 Refseq
Affymetrix Human Clariom D Assay TC1100012535.hg.1 NM_002855.4 Refseq
Affymetrix Human Clariom D Assay TC1100012535.hg.1 NM_002855.4 Refseq
Affymetrix Human Clariom D Assay TC1100012535.hg.1 NM_002855.4 Refseq
Affymetrix Human Clariom D Assay TC1100012535.hg.1 NM_002855.4 Refseq
Affymetrix Human Clariom D Assay TC1100012535.hg.1 NM_002855.4 Refseq
Affymetrix Human Clariom D Assay TC1100012535.hg.1 NM_203285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012535.hg.1 NM_203285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012535.hg.1 NM_203285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012535.hg.1 NM_203285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012535.hg.1 NM_203285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012535.hg.1 NM_203285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012535.hg.1 NM_203285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012535.hg.1 NM_203285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012535.hg.1 NM_203285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012535.hg.1 NM_203285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012535.hg.1 NM_203285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012535.hg.1 NM_203285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012535.hg.1 NM_203285.1 Refseq
Affymetrix Human Clariom D Assay TC1100012535.hg.1 NM_203286.1 Refseq
Affymetrix Human Clariom D Assay TC1100012535.hg.1 NM_203286.1 Refseq
Affymetrix Human Clariom D Assay TC1100012535.hg.1 NM_203286.1 Refseq
Affymetrix Human Clariom D Assay TC1100012535.hg.1 NM_203286.1 Refseq
Affymetrix Human Clariom D Assay TC1100012535.hg.1 NM_203286.1 Refseq
Affymetrix Human Clariom D Assay TC1100012535.hg.1 NM_203286.1 Refseq
Affymetrix Human Clariom D Assay TC1100012535.hg.1 NM_203286.1 Refseq
Affymetrix Human Clariom D Assay TC1100012535.hg.1 NM_203286.1 Refseq
Affymetrix Human Clariom D Assay TC1100012535.hg.1 NM_203286.1 Refseq
Affymetrix Human Clariom D Assay TC1100012535.hg.1 NM_203286.1 Refseq
Affymetrix Human Clariom D Assay PSR1100196705.hg.1 ENST00000341398.6 Ensembl
Affymetrix Human Clariom D Assay PSR1100196705.hg.1 ENST00000531468.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100196705.hg.1 NM_203285.1 Refseq
Affymetrix Human Clariom D Assay PSR1100196705.hg.1 ENST00000341398.6 Ensembl
Affymetrix Human Clariom D Assay PSR1100196705.hg.1 ENST00000531468.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100196705.hg.1 NM_203285.1 Refseq
Affymetrix Human Clariom D Assay PSR1100196705.hg.1 TC1100012535.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1100196706.hg.1 ENST00000341398.6 Ensembl
Affymetrix Human Clariom D Assay PSR1100196706.hg.1 NM_203285.1 Refseq
Affymetrix Human Clariom D Assay PSR1100196706.hg.1 ENST00000341398.6 Ensembl
Affymetrix Human Clariom D Assay PSR1100196706.hg.1 NM_203285.1 Refseq
Affymetrix Human Clariom D Assay PSR1100196706.hg.1 TC1100012535.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-5thf6:80/100.66.79.225:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0