This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01635056_s1
See other TMEM183A GE Assays ›
Gene Symbol:
TMEM183A
Gene Name:
transmembrane protein 183A
Gene Aliases:
C1orf37
Chromosome Location:
Chr.1: 203007367 - 203024848 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01635056_s1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMEM183A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001322955.1 NP_001309884.1 8 2594 104
NM_001322956.1 NP_001309885.1 8 2602 104
NM_001322957.1 NP_001309886.1 8 2597 104
NM_001322958.1 NP_001309887.1 7 2515 104
NM_001322959.1 NP_001309888.1 7 2512 104
NR_136530.1 9 2769 104
NR_136531.1 8 2582 104
NR_136532.1 8 2585 104
NR_136533.1 9 2766 104
NR_136534.1 9 2766 104
GenBank mRNA AL133052.1 - 1 1056 104

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Gene Ontology Categories:

Process(es)

protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000463015.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000463015.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000463015.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000468449.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000468449.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000468449.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000468449.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000468449.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000468449.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000468449.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000488097.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000488097.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000488097.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000488097.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000488097.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000543891.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000543891.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000543891.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000543891.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000543891.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000543891.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000543891.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 ENST00000543891.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000367242.3 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000463015.1 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000463015.1 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000463015.1 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000468449.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000468449.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000468449.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000468449.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000468449.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000468449.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000468449.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000488097.2 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000488097.2 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000488097.2 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000488097.2 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000488097.2 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000543891.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000543891.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000543891.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000543891.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000543891.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000543891.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000543891.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 ENST00000543891.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay TC0100011242.hg.1 NM_138391.5 Refseq
Affymetrix Human Clariom D Assay TC0100011242.hg.1 NM_138391.5 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0