This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01650896_m1
See other UBE2D3 GE Assays ›
Gene Symbol:
UBE2D3
Gene Name:
ubiquitin conjugating enzyme E2 D3
Gene Aliases:
E2(17)KB3, UBC4/5, UBCH5C
Chromosome Location:
Chr.4: 102794383 - 102868893 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01650896_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UBE2D3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_181888.3 NP_871617.1 1 - 2 216 76
GenBank mRNA DA335600.1 - 1 - 2 117 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0400011466.hg.1
This assay targets the same location as HTA transcript cluster(s) TC04001431.hg.1

Additional Information:

This assay is in the 5'UTR region of the NM_181888.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene UBE2D3

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
protein polyubiquitination
DNA repair
cellular protein modification process
ubiquitin-dependent protein catabolic process
protein monoubiquitination
apoptotic process
protein ubiquitination
BMP signaling pathway
TRIF-dependent toll-like receptor signaling pathway
proteasome-mediated ubiquitin-dependent protein catabolic process
protein autoubiquitination
regulation of transcription from RNA polymerase II promoter in response to hypoxia
protein K48-linked ubiquitination
protein K11-linked ubiquitination
positive regulation of protein targeting to mitochondrion
ubiquitin-protein transferase activity
protein binding
ATP binding
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline