This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01650937_m1
See other TMPRSS4 GE Assays ›
Gene Symbol:
TMPRSS4
Gene Name:
transmembrane protease, serine 4
Gene Aliases:
CAPH2, MT-SP2, TMPRSS3
Chromosome Location:
Chr.11: 118077012 - 118125505 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01650937_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMPRSS4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110734.1 10 - 11 1293 88
GenBank mRNA AK290361.1 - 10 - 11 1227 88
BC004855.1 - 10 - 11 1244 3835263 88

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
receptor-mediated endocytosis
serine-type endopeptidase activity
scavenger receptor activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-dd86746ff-8rs9g:80/100.66.75.30:80.
git-commit: 04e44c58f20fafe66519586ffe6db5d8a90e52ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.53.1-Offline