This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01662547_m1
See other FABP5P3 GE Assays ›
Gene Symbol:
FABP5P3
Gene Name:
fatty acid binding protein 5 pseudogene 3
Gene Aliases:
FABP5L3
Chromosome Location:
Chr.7: 152436895 - 152443016 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01662547_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FABP5P3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_002935.1 1 - 2 137 98

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700009737.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07001026.hg.1

Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

transport
transporter activity
lipid binding

MicroArray Probes:

Agilent Affymetrix ArrayStar ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Human CGH Microarray 4x180K 75355 - -
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 21717 - -
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 660621 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 98837 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 109113 BC026342.1 genbank_mrna_human
Agilent SurePrint G3 Human CNV Microarray 2x400K 109113 BC037333.1 genbank_mrna_human
Agilent SurePrint G3 Human CNV Microarray 2x400K 109113 BC078163.1 genbank_mrna_human
Agilent SurePrint G3 Human CNV Microarray 2x400K 109113 CR597272.1 genbank_mrna_human
Agilent SurePrint G3 Human CNV Microarray 2x400K 109113 CR607632.1 genbank_mrna_human
Agilent SurePrint G3 Human CNV Microarray 2x400K 109113 CR624484.1 genbank_mrna_human
Agilent SurePrint G3 Human CNV Microarray 2x400K 109113 J03260.1 genbank_mrna_human
Agilent SurePrint G3 Human CNV Microarray 2x400K 109113 NM_002073.2 refseq_mrna_human_NM
Agilent SurePrint G3 Human CNV Microarray 2x400K 275253 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 84317 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_3938 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_3939 - -
Agilent Human Genome CGH + SNP Microarray A_18_P16332251 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_24_P673063 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26142173 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16040229 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16040217 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26142177 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875817 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16040213 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875818 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26142210 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16040226 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875815 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26142173 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26142177 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875818 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16040213 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875817 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16040217 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05875815 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26142210 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16040229 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16040226 - -
Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P16332251 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.0 ST Array 8147056 - -
Affymetrix Human Gene 1.0 ST Array 7948421 - -
Affymetrix Human Gene 1.0 ST Array 8147054 - -
Affymetrix Human Gene 1.0 ST Array 8147052 - -
Affymetrix Human Gene 1.0 ST Array 8147053 - -
Affymetrix Human Gene 1.0 ST Array 7948422 - -
Affymetrix Human Gene 1.1 ST Array 7948420 - -
Affymetrix Human Gene 1.1 ST Array 8147049 - -
Affymetrix Human Transcriptome Array 2.0 TC0700009737.hg.1 ENST00000437960.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009737.hg.1 ENST00000477993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009737.hg.1 ENST00000477993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009737.hg.1 ENST00000477993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009737.hg.1 ENST00000477993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009737.hg.1 ENST00000477993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009737.hg.1 NR_002935.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009737.hg.1 NR_002935.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009737.hg.1 NR_002935.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009737.hg.1 NR_002935.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07015921.hg.1 TC07001026.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR07015921.hg.1 ENST00000477993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07015921.hg.1 NR_002935.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07015922.hg.1 TC07001026.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR07015922.hg.1 ENST00000477993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07015922.hg.1 NR_002935.1 Refseq
Affymetrix Human Clariom D Assay PSR0700169508.hg.1 ENST00000477993.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700169508.hg.1 NR_002935.1 Refseq
Affymetrix Human Clariom D Assay PSR0700169508.hg.1 ENST00000477993.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700169508.hg.1 NR_002935.1 Refseq
Affymetrix Human Clariom D Assay PSR0700169508.hg.1 TC0700009737.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0700009737.hg.1 ENST00000437960.1 Ensembl
Affymetrix Human Clariom D Assay TC0700009737.hg.1 ENST00000477993.1 Ensembl
Affymetrix Human Clariom D Assay TC0700009737.hg.1 ENST00000477993.1 Ensembl
Affymetrix Human Clariom D Assay TC0700009737.hg.1 ENST00000477993.1 Ensembl
Affymetrix Human Clariom D Assay TC0700009737.hg.1 ENST00000477993.1 Ensembl
Affymetrix Human Clariom D Assay TC0700009737.hg.1 ENST00000477993.1 Ensembl
Affymetrix Human Clariom D Assay TC0700009737.hg.1 NR_002935.1 Refseq
Affymetrix Human Clariom D Assay TC0700009737.hg.1 NR_002935.1 Refseq
Affymetrix Human Clariom D Assay TC0700009737.hg.1 NR_002935.1 Refseq
Affymetrix Human Clariom D Assay TC0700009737.hg.1 NR_002935.1 Refseq
Affymetrix Human Clariom D Assay PSR0700169511.hg.1 ENST00000477993.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700169511.hg.1 NR_002935.1 Refseq
Affymetrix Human Clariom D Assay PSR0700169511.hg.1 ENST00000477993.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700169511.hg.1 NR_002935.1 Refseq
Affymetrix Human Clariom D Assay PSR0700169511.hg.1 TC0700009737.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0700169510.hg.1 ENST00000437960.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700169510.hg.1 ENST00000477993.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700169510.hg.1 NR_002935.1 Refseq
Affymetrix Human Clariom D Assay PSR0700169510.hg.1 ENST00000437960.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700169510.hg.1 ENST00000477993.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700169510.hg.1 NR_002935.1 Refseq
Affymetrix Human Clariom D Assay PSR0700169510.hg.1 TC0700009737.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0700169509.hg.1 ENST00000477993.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700169509.hg.1 NR_002935.1 Refseq
Affymetrix Human Clariom D Assay PSR0700169509.hg.1 ENST00000477993.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700169509.hg.1 NR_002935.1 Refseq
Affymetrix Human Clariom D Assay PSR0700169509.hg.1 TC0700009737.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
Arraystar Human LncRNA microarray V2.0 ASHG19A3A035362 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A035362 - -
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 101498 - -
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_2217574 NR_002935.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_2217574 CR621082.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_2217574 NR_002935.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_2217574 CR621082.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2217574 NR_002935.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2217574 CR621082.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2217574 NR_002935.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2217574 CR621082.1 human_transcriptome
Illumina HumanRef-8 v3.0 expression beadchip ILMN_2217574 NR_002935.1 human_transcriptome
Illumina HumanRef-8 v3.0 expression beadchip ILMN_2217574 CR621082.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_2217574 NR_002935.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_2217574 CR621082.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_2217574 NR_002935.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_2217574 CR621082.1 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_2217574 NR_002935.1 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_2217574 CR621082.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline