This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01673243_m1
See other STARD4 GE Assays ›
Gene Symbol:
STARD4
Gene Name:
StAR related lipid transfer domain containing 4
Gene Aliases:
-
Chromosome Location:
Chr.5: 111496033 - 111512597 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01673243_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: STARD4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_131753.1 2 - 3 319 126
NR_131754.1 2 - 3 313 126
XM_017009041.1 XP_016864530.1 2 - 3 257 126
XM_017009042.1 XP_016864531.1 2 - 3 251 126
GenBank mRNA AK125317.1 - 2 - 3 251 126
AK310848.1 - 2 - 3 243 126
AK315869.1 - 2 - 3 257 126
BC019632.1 - 2 - 3 245 126

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK315869.1, XM_017009041.1, XM_017009042.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

lipid transport
positive regulation of cholesterol esterification
positive regulation of bile acid biosynthetic process
cholesterol binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline