This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01676444_g1
See other RAC2 GE Assays ›
Gene Symbol:
RAC2
Gene Name:
ras-related C3 botulinum toxin substrate 2 (rho family, small GTP binding protein Rac2)
Gene Aliases:
EN-7, Gx, HSPC022, p21-Rac2
Chromosome Location:
Chr.22: 37225261 - 37244299 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01676444_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RAC2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK096924.1 - 2 - 3 214 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000249071.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000249071.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000249071.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000249071.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000249071.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000249071.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000249071.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000249071.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000249071.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000249071.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000249071.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000249071.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000401529.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000401529.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000401529.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000401529.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000405484.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000405484.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000405484.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000405484.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000405484.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000405484.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000405484.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000405484.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000405484.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000406508.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000406508.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000406508.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000406508.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000406508.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000406508.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000406508.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000406508.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000406508.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000406508.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000441619.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000441619.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000441619.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000441619.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000441619.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000441619.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000441619.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000469532.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000469532.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000469532.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000469532.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000469532.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000481215.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000481215.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000481215.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000481215.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000481215.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 ENST00000481215.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 NM_002872.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 NM_002872.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 NM_002872.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 NM_002872.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 NM_002872.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 NM_002872.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 NM_002872.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 NM_002872.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 NM_002872.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 NM_002872.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 NM_002872.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008641.hg.1 NM_002872.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-bm6jx:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0