This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01904366_s1
See other ZBTB46-AS1 GE Assays ›
Gene Symbol:
ZBTB46-AS1
Gene Name:
ZBTB46 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.20: 63808076 - 63823015 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01904366_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZBTB46-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110081.1 3 732 137

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2000008092.hg.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR2000156666.hg.1 NR_110081.1 Refseq
Affymetrix Human Clariom D Assay PSR2000156666.hg.1 TC2000008092.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2000156666.hg.1 NR_110081.1 Refseq
Affymetrix Human Clariom D Assay PSR2000156650.hg.1 ENST00000435912.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000156650.hg.1 NR_110081.1 Refseq
Affymetrix Human Clariom D Assay PSR2000156650.hg.1 TC2000008092.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2000156650.hg.1 ENST00000435912.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000156650.hg.1 NR_110081.1 Refseq
Affymetrix Human Clariom D Assay PSR2000156665.hg.1 NR_110081.1 Refseq
Affymetrix Human Clariom D Assay PSR2000156665.hg.1 TC2000008092.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2000156665.hg.1 NR_110081.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0