This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01924130_s1
See other HIST1H3I GE Assays ›
Gene Symbol:
HIST1H3I
Gene Name:
histone cluster 1, H3i
Gene Aliases:
H3.f, H3/f, H3FF
Chromosome Location:
Chr.6: 27871845 - 27872321 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01924130_s1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HIST1H3I
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003533.2 NP_003524.1 1 137 133
GenBank mRNA BC066884.1 - 1 137 7002077 133
BC067493.1 - 1 137 7002079 133
BC069305.1 - 1 137 7262745 133

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600011233.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06001427.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene HIST1H3I

Panther Classification:

Molecular Function -

histone

Gene Ontology Categories:

Function(s) Process(es)

chromatin silencing at rDNA
positive regulation of defense response to virus by host
nucleosome assembly
DNA replication-dependent nucleosome assembly
blood coagulation
gene silencing by RNA
telomere organization
cellular protein metabolic process
negative regulation of gene expression, epigenetic
positive regulation of gene expression, epigenetic
protein heterotetramerization
regulation of gene silencing
cell-cell adhesion
xenophagy
DNA binding
protein binding
histone binding
protein heterodimerization activity
cadherin binding involved in cell-cell adhesion

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR06016361.hg.1 TC06001427.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR06016361.hg.1 ENST00000328488.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06016361.hg.1 NM_003533.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011233.hg.1 ENST00000616365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011233.hg.1 NM_003533.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011233.hg.1 NM_003533.2 Refseq
Affymetrix Human Clariom D Assay TC0600011233.hg.1 ENST00000616365.1 Ensembl
Affymetrix Human Clariom D Assay TC0600011233.hg.1 NM_003533.2 Refseq
Affymetrix Human Clariom D Assay TC0600011233.hg.1 NM_003533.2 Refseq
Affymetrix Human Clariom D Assay PSR0600175736.hg.1 ENST00000616365.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600175736.hg.1 NM_003533.2 Refseq
Affymetrix Human Clariom D Assay PSR0600175736.hg.1 ENST00000616365.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600175736.hg.1 NM_003533.2 Refseq
Affymetrix Human Clariom D Assay PSR0600175736.hg.1 TC0600011233.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0