This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01924151_s1
See other HIST1H4B GE Assays ›
Gene Symbol:
HIST1H4B
Gene Name:
histone cluster 1, H4b
Gene Aliases:
H4/I, H4FI
Chromosome Location:
Chr.6: 26026896 - 26027252 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01924151_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HIST1H4B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003544.2 NP_003535.1 1 305 94
GenBank mRNA BC069467.1 - 1 305 7262716 94

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600011123.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene HIST1H4B

Panther Classification:

Molecular Function -

histone

Gene Ontology Categories:

Function(s) Process(es)

chromatin silencing at rDNA
double-strand break repair via nonhomologous end joining
nucleosome assembly
DNA replication-dependent nucleosome assembly
DNA replication-independent nucleosome assembly
DNA-templated transcription, initiation
telomere capping
gene silencing by RNA
telomere organization
CENP-A containing nucleosome assembly
cellular protein metabolic process
negative regulation of megakaryocyte differentiation
negative regulation of gene expression, epigenetic
positive regulation of gene expression, epigenetic
protein heterotetramerization
beta-catenin-TCF complex assembly
DNA binding
protein binding
protein domain specific binding
histone binding
poly(A) RNA binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600011123.hg.1 ENST00000377745.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011123.hg.1 ENST00000377745.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011123.hg.1 ENST00000377745.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011123.hg.1 NM_003544.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011123.hg.1 NM_003544.2 Refseq
Affymetrix Human Clariom D Assay PSR0600175273.hg.1 ENST00000377745.3 Ensembl
Affymetrix Human Clariom D Assay PSR0600175273.hg.1 NM_003544.2 Refseq
Affymetrix Human Clariom D Assay PSR0600175273.hg.1 ENST00000377745.3 Ensembl
Affymetrix Human Clariom D Assay PSR0600175273.hg.1 NM_003544.2 Refseq
Affymetrix Human Clariom D Assay PSR0600175273.hg.1 TC0600011123.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0600011123.hg.1 ENST00000377745.3 Ensembl
Affymetrix Human Clariom D Assay TC0600011123.hg.1 ENST00000377745.3 Ensembl
Affymetrix Human Clariom D Assay TC0600011123.hg.1 ENST00000377745.3 Ensembl
Affymetrix Human Clariom D Assay TC0600011123.hg.1 NM_003544.2 Refseq
Affymetrix Human Clariom D Assay TC0600011123.hg.1 NM_003544.2 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 132162 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0