This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02339407_m1
See other CD200R1L GE Assays ›
Gene Symbol:
CD200R1L
Gene Name:
CD200 receptor 1 like
Gene Aliases:
CD200R2, CD200RLa
Chromosome Location:
Chr.3: 112814925 - 112847004 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs02339407_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CD200R1L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001008784.2 NP_001008784.2 1 - 2 266 94
GenBank mRNA AY552790.1 - 1 - 2 266 94

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300012006.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03001653.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CD200R1L

Panther Classification:

Molecular Function -

transmembrane signal receptor

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 ENST00000398214.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 ENST00000398214.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 ENST00000398214.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 ENST00000398214.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 ENST00000398214.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 ENST00000398214.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 ENST00000398214.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 ENST00000398214.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 ENST00000448932.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 ENST00000448932.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 ENST00000448932.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 ENST00000448932.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 ENST00000486723.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 ENST00000486723.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 ENST00000486723.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 ENST00000486723.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 ENST00000486723.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 ENST00000486723.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 ENST00000486723.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 ENST00000486723.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 ENST00000486723.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 ENST00000488794.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 ENST00000488794.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 ENST00000488794.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 ENST00000488794.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 ENST00000488794.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 ENST00000488794.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 ENST00000488794.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 ENST00000488794.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 ENST00000488794.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 ENST00000488794.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 ENST00000617549.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 ENST00000617549.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 ENST00000617549.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 ENST00000617549.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 NM_001008784.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 NM_001008784.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 NM_001008784.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 NM_001008784.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 NM_001008784.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 NM_001008784.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 NM_001008784.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 NM_001008784.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 NM_001199215.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 NM_001199215.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 NM_001199215.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 NM_001199215.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 NM_001199215.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 NM_001199215.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 NM_001199215.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 NM_001199215.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012006.hg.1 NM_001199215.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03029801.hg.1 ENST00000398214.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03029801.hg.1 ENST00000448932.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03029801.hg.1 ENST00000488794.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03029801.hg.1 NM_001008784.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03029801.hg.1 NM_001199215.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03029801.hg.1 TC03001653.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03029804.hg.1 ENST00000398214.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03029804.hg.1 ENST00000448932.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03029804.hg.1 ENST00000488794.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03029804.hg.1 NM_001008784.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03029804.hg.1 NM_001199215.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03029804.hg.1 TC03001653.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03029805.hg.1 ENST00000398214.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03029805.hg.1 ENST00000448932.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03029805.hg.1 ENST00000488794.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03029805.hg.1 NM_001008784.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03029805.hg.1 NM_001199215.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03029805.hg.1 TC03001653.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0300012006.hg.1 ENST00000398214.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012006.hg.1 ENST00000398214.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012006.hg.1 ENST00000398214.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012006.hg.1 ENST00000398214.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012006.hg.1 ENST00000398214.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012006.hg.1 ENST00000398214.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012006.hg.1 ENST00000398214.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012006.hg.1 ENST00000398214.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012006.hg.1 ENST00000448932.4 Ensembl
Affymetrix Human Clariom D Assay TC0300012006.hg.1 ENST00000448932.4 Ensembl
Affymetrix Human Clariom D Assay TC0300012006.hg.1 ENST00000448932.4 Ensembl
Affymetrix Human Clariom D Assay TC0300012006.hg.1 ENST00000448932.4 Ensembl
Affymetrix Human Clariom D Assay TC0300012006.hg.1 ENST00000486723.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012006.hg.1 ENST00000486723.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012006.hg.1 ENST00000486723.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012006.hg.1 ENST00000486723.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012006.hg.1 ENST00000486723.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012006.hg.1 ENST00000486723.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012006.hg.1 ENST00000486723.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012006.hg.1 ENST00000486723.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012006.hg.1 ENST00000486723.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012006.hg.1 ENST00000488794.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012006.hg.1 ENST00000488794.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012006.hg.1 ENST00000488794.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012006.hg.1 ENST00000488794.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012006.hg.1 ENST00000488794.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012006.hg.1 ENST00000488794.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012006.hg.1 ENST00000488794.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012006.hg.1 ENST00000488794.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012006.hg.1 ENST00000488794.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012006.hg.1 ENST00000488794.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012006.hg.1 ENST00000617549.3 Ensembl
Affymetrix Human Clariom D Assay TC0300012006.hg.1 ENST00000617549.3 Ensembl
Affymetrix Human Clariom D Assay TC0300012006.hg.1 ENST00000617549.3 Ensembl
Affymetrix Human Clariom D Assay TC0300012006.hg.1 ENST00000617549.3 Ensembl
Affymetrix Human Clariom D Assay TC0300012006.hg.1 NM_001008784.2 Refseq
Affymetrix Human Clariom D Assay TC0300012006.hg.1 NM_001008784.2 Refseq
Affymetrix Human Clariom D Assay TC0300012006.hg.1 NM_001008784.2 Refseq
Affymetrix Human Clariom D Assay TC0300012006.hg.1 NM_001008784.2 Refseq
Affymetrix Human Clariom D Assay TC0300012006.hg.1 NM_001008784.2 Refseq
Affymetrix Human Clariom D Assay TC0300012006.hg.1 NM_001008784.2 Refseq
Affymetrix Human Clariom D Assay TC0300012006.hg.1 NM_001008784.2 Refseq
Affymetrix Human Clariom D Assay TC0300012006.hg.1 NM_001008784.2 Refseq
Affymetrix Human Clariom D Assay TC0300012006.hg.1 NM_001199215.1 Refseq
Affymetrix Human Clariom D Assay TC0300012006.hg.1 NM_001199215.1 Refseq
Affymetrix Human Clariom D Assay TC0300012006.hg.1 NM_001199215.1 Refseq
Affymetrix Human Clariom D Assay TC0300012006.hg.1 NM_001199215.1 Refseq
Affymetrix Human Clariom D Assay TC0300012006.hg.1 NM_001199215.1 Refseq
Affymetrix Human Clariom D Assay TC0300012006.hg.1 NM_001199215.1 Refseq
Affymetrix Human Clariom D Assay TC0300012006.hg.1 NM_001199215.1 Refseq
Affymetrix Human Clariom D Assay TC0300012006.hg.1 NM_001199215.1 Refseq
Affymetrix Human Clariom D Assay TC0300012006.hg.1 NM_001199215.1 Refseq
Affymetrix Human Clariom D Assay PSR0300189379.hg.1 ENST00000398214.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300189379.hg.1 ENST00000448932.4 Ensembl
Affymetrix Human Clariom D Assay PSR0300189379.hg.1 ENST00000486723.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300189379.hg.1 ENST00000488794.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300189379.hg.1 ENST00000617549.3 Ensembl
Affymetrix Human Clariom D Assay PSR0300189379.hg.1 NM_001008784.2 Refseq
Affymetrix Human Clariom D Assay PSR0300189379.hg.1 NM_001199215.1 Refseq
Affymetrix Human Clariom D Assay PSR0300189379.hg.1 ENST00000398214.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300189379.hg.1 ENST00000448932.4 Ensembl
Affymetrix Human Clariom D Assay PSR0300189379.hg.1 ENST00000486723.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300189379.hg.1 ENST00000488794.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300189379.hg.1 ENST00000617549.3 Ensembl
Affymetrix Human Clariom D Assay PSR0300189379.hg.1 NM_001008784.2 Refseq
Affymetrix Human Clariom D Assay PSR0300189379.hg.1 NM_001199215.1 Refseq
Affymetrix Human Clariom D Assay PSR0300189379.hg.1 TC0300012006.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300189384.hg.1 ENST00000398214.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300189384.hg.1 ENST00000486723.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300189384.hg.1 ENST00000488794.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300189384.hg.1 NM_001008784.2 Refseq
Affymetrix Human Clariom D Assay PSR0300189384.hg.1 NM_001199215.1 Refseq
Affymetrix Human Clariom D Assay PSR0300189384.hg.1 ENST00000398214.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300189384.hg.1 ENST00000486723.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300189384.hg.1 ENST00000488794.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300189384.hg.1 NM_001008784.2 Refseq
Affymetrix Human Clariom D Assay PSR0300189384.hg.1 NM_001199215.1 Refseq
Affymetrix Human Clariom D Assay PSR0300189384.hg.1 TC0300012006.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300189385.hg.1 ENST00000398214.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300189385.hg.1 ENST00000486723.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300189385.hg.1 ENST00000488794.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300189385.hg.1 NM_001008784.2 Refseq
Affymetrix Human Clariom D Assay PSR0300189385.hg.1 NM_001199215.1 Refseq
Affymetrix Human Clariom D Assay PSR0300189385.hg.1 ENST00000398214.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300189385.hg.1 ENST00000486723.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300189385.hg.1 ENST00000488794.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300189385.hg.1 NM_001008784.2 Refseq
Affymetrix Human Clariom D Assay PSR0300189385.hg.1 NM_001199215.1 Refseq
Affymetrix Human Clariom D Assay PSR0300189385.hg.1 TC0300012006.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline