This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02383055_s1
See other SLC7A5 GE Assays ›
Gene Symbol:
SLC7A5
Gene Name:
solute carrier family 7 member 5
Gene Aliases:
4F2LC, CD98, D16S469E, E16, LAT1, MPE16, hLAT1
Chromosome Location:
Chr.16: 87830022 - 87869499 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs02383055_s1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC7A5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003486.5 NP_003477.4
XM_006721286.3 XP_006721349.2
GenBank mRNA AB018009.1 -
AF104032.1 -
BC039692.2 - 5551612
BC042600.1 - 4470593
M80244.1 -

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

cellular amino acid metabolic process
transport
amino acid transport
nervous system development
neutral amino acid transport
L-amino acid transport
cell differentiation
leukocyte migration
L-alpha-amino acid transmembrane transport
amino acid transmembrane transporter activity
neutral amino acid transmembrane transporter activity
L-amino acid transmembrane transporter activity
antiporter activity
peptide antigen binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline