This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02385033_g1
See other RPL6 GE Assays ›
Gene Symbol:
RPL6 RPL6P27
Gene Name:
ribosomal protein L6
ribosomal protein L6 pseudogene 27
Gene Aliases:
L6, SHUJUN-2, TAXREB107, TXREB1
RPL6_10_1582
Chromosome Location:
Chr.12: 112405181 - 112418850 on Build GRCh38
Chromosome Location:
Chr.18: 6462110 - 6463029 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
-
Assay ID Hs02385033_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPL6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000970.4 NP_000961.2 3 - 4 391 104
NM_001024662.2 NP_001019833.1 3 - 4 567 104
NM_001320137.1 NP_001307066.1 3 - 4 572 104
NM_001320138.1 NP_001307067.1 3 - 4 606 104
NM_001320139.1 NP_001307068.1 3 - 4 453 104
NM_001320140.1 NP_001307069.1 3 - 4 615 104
NM_001320141.1 NP_001307070.1 5 - 6 680 104
NM_001320142.1 NP_001307071.1 3 - 4 327 104
XM_017019781.1 XP_016875270.1 5 - 6 1715 104
XM_017019782.1 XP_016875271.1 5 - 6 876 104
GenBank mRNA AF261087.1 - 1 358 104
AK299470.1 - 4 - 5 104
AK311863.1 - 3 - 4 355 104
BC004138.2 - 1 367 2823733 104
BC013863.1 - 1 341 104
BC020679.1 - 1 355 4616668 104
BC022444.1 - 3 - 4 522 4286423 104
BC031009.1 - 1 355 4333761 104
BC032299.1 - 1 352 4994660 104
BC071912.1 - 1 344 6293404 104
BC104824.1 - 3 - 4 367 8143827 104
BC104826.1 - 3 - 4 367 8143829 104
BG030827.1 - 1 117 104
BP285025.1 - 3 - 4 391 104
CB140996.1 - 3 - 4 520 104
CD049618.1 - 3 - 4 537 104
D17554.1 - 3 - 4 357 104
S71022.1 - 2 - 3 201 104
X69391.1 - 3 - 4 362 104
Gene Symbol: RPL6P27
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK126570.1 - 1102 105

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001320142.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RPL6
Click here to find siRNAs for Gene RPL6P27

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

ribosomal large subunit assembly
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
cytoplasmic translation
regulation of transcription, DNA-templated
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
cell-cell adhesion
DNA binding
RNA binding
structural constituent of ribosome
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline