This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02568337_s1
See other GPSM1 GE Assays ›
Gene Symbol:
GPSM1
Gene Name:
G-protein signaling modulator 1
Gene Aliases:
AGS3
Chromosome Location:
Chr.9: 136327476 - 136359605 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs02568337_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPSM1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001145638.2 NP_001139110.2 13 1918 111
NM_001145639.1 NP_001139111.1 3 342 111
NM_001200003.1 NP_001186932.1 3 466 111
XM_011518498.2 XP_011516800.1 13 1809 111
XM_017014598.1 XP_016870087.1 12 1717 111
XM_017014599.1 XP_016870088.1 5 684 111
GenBank mRNA AL117478.1 - 7 758 111
AL545250.3 - 3 466 111
AY173053.1 - 12 1629 111
BC048343.1 - 2 403 111
BI766024.1 - 3 373 111

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

nervous system development
regulation of G-protein coupled receptor protein signaling pathway
cell differentiation
regulation of catalytic activity
GDP-dissociation inhibitor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0