This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02568404_s1
See other TRNT1 GE Assays ›
Gene Symbol:
TRNT1
Gene Name:
tRNA nucleotidyl transferase 1
Gene Aliases:
CCA1, CGI-47, MtCCA, RPEM, SIFD
Chromosome Location:
Chr.3: 3126916 - 3153435 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs02568404_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TRNT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001302946.1 NP_001289875.1 4 506 72
NM_182916.2 NP_886552.2 4 506 72
XM_011533776.2 XP_011532078.1 4 504 72
XM_011533777.2 XP_011532079.1 4 752 72
XM_011533778.2 XP_011532080.1 4 506 72
GenBank mRNA AB063105.1 - 4 449 72
AF151805.1 - 4 448 72
AK290411.1 - 4 471 72
AL834397.1 - 4 470 72
BC012537.1 - 4 457 4097092 72
DB032432.1 - 4 507 72

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TRNT1

Panther Classification:

Molecular Function -

RNA metabolism protein RNA processing factor nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

tRNA 3'-terminal CCA addition
protein targeting to mitochondrion
tRNA 3'-end processing
mitochondrial tRNA 3'-end processing
tRNA binding
tRNA adenylyltransferase activity
ATP binding
RNA polymerase activity
CTP:tRNA cytidylyltransferase activity
CTP:3'-cytidine-tRNA cytidylyltransferase activity
ATP:3'-cytidine-cytidine-tRNA adenylyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0