This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02569955_s1
See other COTL1 GE Assays ›
Gene Symbol:
COTL1
Gene Name:
coactosin like F-actin binding protein 1
Gene Aliases:
CLP
Chromosome Location:
Chr.16: 84565596 - 84618096 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs02569955_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: COTL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021149.3 NP_066972.1 3 390 63
GenBank mRNA AK026033.1 - 2 115 63
AK315675.1 - 3 369 63
BC010039.2 - 3 357 3604770 63
BC010884.1 - 3 306 3537437 63
BC016702.1 - 3 375 4068379 63
BC042970.1 - 3 377 5288618 63
BC053682.1 - 3 353 5113263 63
BT006968.1 - 3 208 63
DA968872.1 - 3 378 63
L54057.1 - 3 357 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600011060.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene COTL1

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein cytoskeletal protein non-motor actin binding protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
defense response to fungus
actin binding
protein binding
enzyme binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000561707.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000561707.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000561707.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000561707.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000564057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000564057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000564057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000564057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000564057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000564057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000564057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000564057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000564662.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000564662.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000564662.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000564662.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000564662.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000564662.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000567278.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000567278.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000567278.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000567278.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000567278.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000567278.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000567278.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000567278.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000567786.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000567786.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000561707.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000561707.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000561707.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000561707.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000564057.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000564057.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000564057.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000564057.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000564057.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000564057.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000564057.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000564057.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000564662.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000564662.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000564662.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000564662.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000564662.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000564662.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000567278.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000567278.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000567278.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000567278.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000567278.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000567278.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000567278.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000567278.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000567786.2 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000567786.2 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Clariom D Assay TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Clariom D Assay TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Clariom D Assay TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Clariom D Assay TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Clariom D Assay TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Clariom D Assay TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Clariom D Assay TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Clariom D Assay TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Clariom D Assay TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Clariom D Assay PSR1600186000.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Clariom D Assay PSR1600186000.hg.1 ENST00000561707.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600186000.hg.1 ENST00000564057.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600186000.hg.1 ENST00000564662.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600186000.hg.1 NM_021149.3 Refseq
Affymetrix Human Clariom D Assay PSR1600186000.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Clariom D Assay PSR1600186000.hg.1 ENST00000561707.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600186000.hg.1 ENST00000564057.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600186000.hg.1 ENST00000564662.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600186000.hg.1 NM_021149.3 Refseq
Affymetrix Human Clariom D Assay PSR1600186000.hg.1 TC1600011060.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0