This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02597447_g1
See other NDUFAF5 GE Assays ›
Gene Symbol:
NDUFAF5
Gene Name:
NADH:ubiquinone oxidoreductase complex assembly factor 5
Gene Aliases:
C20orf7, bA526K24.2, dJ842G6.1
Chromosome Location:
Chr.20: 13785026 - 13821580 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs02597447_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NDUFAF5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039375.2 NP_001034464.1 9 - 10 898 94
NM_024120.4 NP_077025.2 10 - 11 982 94
NR_029377.1 11 - 12 1080 94
XM_006723622.2 XP_006723685.1 9 - 10 863 94
XM_006723623.2 XP_006723686.1 9 - 10 995 94
XM_006723624.2 XP_006723687.1 9 - 10 1009 94
XM_011529344.2 XP_011527646.1 7 - 8 1337 94
XM_017028061.1 XP_016883550.1 10 - 11 1094 94
GenBank mRNA AA252680.1 - 1 - 2 94
AA789132.1 - 1 - 2 94
AK025977.1 - 4 - 5 408 94
AK091060.1 - 11 - 12 1057 94
AK289781.1 - 10 - 11 974 94
BC005984.1 - 10 - 11 792 94
BC073158.1 - 9 - 10 875 6053315 94
BM993545.1 - 1 - 2 94

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NDUFAF5

Gene Ontology Categories:

Function(s) Process(es)

methylation
mitochondrial respiratory chain complex I assembly
molecular_function
methyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-fcf5f9d6d-l2zpb:80/100.66.75.138:80.
git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.51.0-2026.05.21-1.0