This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02597744_m1
See other NDUFAF5 GE Assays ›
Gene Symbol:
NDUFAF5
Gene Name:
NADH:ubiquinone oxidoreductase complex assembly factor 5
Gene Aliases:
C20orf7, bA526K24.2, dJ842G6.1
Chromosome Location:
Chr.20: 13785026 - 13821580 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs02597744_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NDUFAF5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_024120.4 NP_077025.2 6 - 7 560 90
NR_029377.1 6 - 7 560 90
XM_006723620.3 XP_006723683.1 6 - 7 537 90
XM_011529342.2 XP_011527644.1 6 - 7 537 90
GenBank mRNA AK091060.1 - 6 - 7 537 90
AK289781.1 - 6 - 7 552 90
BC005984.1 - 6 - 7 370 90

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC005984.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NDUFAF5

Gene Ontology Categories:

Function(s) Process(es)

methylation
mitochondrial respiratory chain complex I assembly
molecular_function
methyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-fcf5f9d6d-9k2pj:80/100.66.79.197:80.
git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.51.0-2026.05.21-1.0