This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02655142_m1
See other CXXC5 GE Assays ›
Gene Symbol:
CXXC5
Gene Name:
CXXC finger protein 5
Gene Aliases:
CF5, HSPC195, RINF, WID
Chromosome Location:
Chr.5: 139647299 - 139683885 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs02655142_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CXXC5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001317199.1 NP_001304128.1 3 - 4 1730 94
NM_001317200.1 NP_001304129.1 2 - 3 1167 94
NM_001317201.1 NP_001304130.1 2 - 3 1219 94
NM_001317202.1 NP_001304131.1 2 - 3 1183 94
NM_001317203.1 NP_001304132.1 3 - 4 1391 94
NM_001317204.1 NP_001304133.1 3 - 4 1261 94
NM_001317205.1 NP_001304134.1 2 - 3 1260 94
NM_001317206.1 NP_001304135.1 4 - 5 1487 94
NM_001317207.1 NP_001304136.1 2 - 3 1169 94
NM_001317208.1 NP_001304137.1 3 - 4 1275 94
NM_001317209.1 NP_001304138.1 2 - 3 1181 94
NM_001317210.1 NP_001304139.1 3 - 4 1269 94
NM_001317211.1 NP_001304140.1 3 - 4 1263 94
NM_016463.8 NP_057547.5 2 - 3 1636 94
XM_017009573.1 XP_016865062.1 2 - 3 1506 94
GenBank mRNA AB097005.1 - 2 - 3 1213 94
AB097032.1 - 2 - 3 1213 94
AF151029.1 - 1 - 2 842 94
AK001782.1 - 2 - 3 1158 94
AK024338.1 - 2 - 3 1636 94
AK314420.1 - 1 - 2 920 94
AY007103.1 - 1 - 2 1281 94
BC002490.2 - 1 - 2 972 3050465 94
BC006428.1 - 2 - 3 1567 3343683 94
BC017439.1 - 2 - 3 1528 2989328 94
BC024040.1 - 2 - 3 1557 3925666 94
BC041013.1 - 2 - 3 1557 4139961 94
BC050046.1 - 2 - 3 1529 94
GQ379202.1 - 1 - 2 922 94

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CXXC5

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
signal transduction
positive regulation of I-kappaB kinase/NF-kappaB signaling
signal transducer activity
protein binding
transcription factor binding
zinc ion binding
sequence-specific DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0