This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02693885_m1
See other DUSP13 GE Assays ›
Gene Symbol:
DUSP13
Gene Name:
dual specificity phosphatase 13
Gene Aliases:
BEDP, DUSP13A, DUSP13B, MDSP, SKRP4, TMDP
Chromosome Location:
Chr.10: 75094432 - 75109221 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs02693885_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DUSP13
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001007272.1 NP_001007273.1 1 - 2 202 65

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DUSP13

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
meiotic nuclear division
spermatogenesis
dephosphorylation
peptidyl-tyrosine dephosphorylation
protein tyrosine phosphatase activity
protein binding
protein tyrosine/serine/threonine phosphatase activity
phosphatase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0