This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02760627_g1
See other RBFOX3 GE Assays ›
Gene Symbol:
RBFOX3
Gene Name:
RNA binding protein, fox-1 homolog 3
Gene Aliases:
FOX-3, FOX3, HRNBP3, NEUN
Chromosome Location:
Chr.17: 79089345 - 79611101 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs02760627_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RBFOX3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK126788.1 - 2 130
BC140939.1 - 1 2012 9056751 130

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1700011941.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3329632 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 38805 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.0 ST Array 8019753 - -
Affymetrix Human Gene 1.0 ST Array 8019752 - -
Affymetrix Human Gene 1.1 ST Array 8019751 - -
Affymetrix Human Transcriptome Array 2.0 TC1700011941.hg.1 ENST00000571452.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011941.hg.1 ENST00000571452.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011941.hg.1 ENST00000571452.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011941.hg.1 ENST00000571452.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011941.hg.1 ENST00000611949.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700197560.hg.1 ENST00000571452.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700197560.hg.1 ENST00000611949.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700197560.hg.1 ENST00000571452.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700197560.hg.1 ENST00000611949.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700197560.hg.1 TC1700011941.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1700011941.hg.1 ENST00000571452.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011941.hg.1 ENST00000571452.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011941.hg.1 ENST00000571452.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011941.hg.1 ENST00000571452.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011941.hg.1 ENST00000611949.1 Ensembl
Array Name Probe IDs Targets Target Type
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1765792 - -
Illumina human-6 v2.0 expression beadchip ILMN_1765792 - -
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1669609 AK126788.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_1669609 AK126788.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0