This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02793376_m1
See other PHACTR3 GE Assays ›
Gene Symbol:
PHACTR3
Gene Name:
phosphatase and actin regulator 3
Gene Aliases:
C20orf101, H17739, PPP1R123, SCAPIN1, SCAPININ
Chromosome Location:
Chr.20: 59577497 - 59849829 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs02793376_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PHACTR3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001199505.1 NP_001186434.1 7 - 8 1164 64
NM_001199506.1 NP_001186435.1 7 - 8 1159 64
NM_001281507.1 NP_001268436.1 7 - 8 1263 64
NM_080672.4 NP_542403.1 7 - 8 1641 64
NM_183244.1 NP_899067.1 7 - 8 1284 64
NM_183246.1 NP_899069.1 6 - 7 1074 64
XM_011528525.2 XP_011526827.1 7 - 8 1270 64
XM_011528526.2 XP_011526828.1 6 - 7 1076 64
XM_017027626.1 XP_016883115.1 7 - 8 1373 64
XM_017027627.1 XP_016883116.1 7 - 8 1318 64
XM_017027628.1 XP_016883117.1 6 - 7 966 64
XM_017027629.1 XP_016883118.1 6 - 7 1109 64
XM_017027630.1 XP_016883119.1 6 - 7 937 64
XM_017027631.1 XP_016883120.1 7 - 8 1200 64
GenBank mRNA AJ311122.2 - 7 - 8 1640 64
AK091632.1 - 7 - 8 1286 64
AK098788.1 - 7 - 8 1164 64
AK295459.1 - 7 - 8 1263 64
AK314493.1 - 7 - 8 1371 64
AK316047.1 - 7 - 8 1156 64
BC108303.1 - 7 - 8 1641 4179728 64
BC117362.1 - 7 - 8 1529 40125913 64
BC117364.1 - 7 - 8 1532 40125915 64

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PHACTR3

Panther Classification:

Molecular Function -

phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of catalytic activity
actin binding
protein phosphatase inhibitor activity
protein phosphatase 1 binding
protein phosphatase type 1 regulator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w64qk:80/100.66.78.26:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0