This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02794119_m1
See other CREM GE Assays ›
Gene Symbol:
CREM
Gene Name:
cAMP responsive element modulator
Gene Aliases:
CREM-2, ICER, hCREM-2
Chromosome Location:
Chr.10: 35126841 - 35212958 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs02794119_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CREM
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001267568.1 NP_001254497.1 1 - 2 89 64
NM_182718.1 NP_874387.1 1 - 2 89 64
NM_182720.1 NP_874389.1 1 - 2 89 64
GenBank mRNA AK290612.1 - 1 - 2 83 64
CD639591.1 - 1 - 2 86 64

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK290612.1, NM_001267568.1, NM_182718.1, NM_182720.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Inventoried Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

glucose metabolic process
transcription, DNA-templated
regulation of transcription, DNA-templated
fatty acid metabolic process
glycosphingolipid metabolic process
signal transduction
multicellular organism development
spermatogenesis
cell differentiation
circadian regulation of gene expression
regulation of circadian rhythm
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
retinoic acid receptor signaling pathway
response to cAMP
core promoter sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
cAMP response element binding protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0