This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02809338_g1
See other TMA7 GE Assays ›
Gene Symbol:
TMA7
Gene Name:
translation machinery associated 7 homolog
Gene Aliases:
CCDC72, HSPC016
Chromosome Location:
Chr.3: 48440276 - 48444129 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs02809338_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMA7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AF077202.1 - 3 - 4 205 102
AF161448.1 - 2 - 3 154 102
AL712164.1 - 3 - 4 182 102
AY508979.1 - 1 188 102
AY794023.1 - 3 - 4 189 102
BC001102.2 - 1 177 102
BC080531.1 - 3 - 4 190 4875991 102
BU536461.1 - 3 - 4 234 102

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-r7bw6:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0