This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02809597_g1
See other EEF1G GE Assays ›
Gene Symbol:
EEF1G
Gene Name:
eukaryotic translation elongation factor 1 gamma
Gene Aliases:
EF1G, GIG35
Chromosome Location:
Chr.11: 62559601 - 62573988 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs02809597_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EEF1G
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001404.4 NP_001395.1 4 - 5 523 66
GenBank mRNA AK092787.1 - 3 - 4 1483 66
AK129569.1 - 4 - 5 396 66
AK129618.1 - 4 - 5 396 66
AK130026.1 - 4 - 5 398 66
AK299876.1 - 4 - 5 395 66
AK300203.1 - 4 - 5 618 66
AK300686.1 - 4 - 5 398 66
AK300729.1 - 4 - 5 328 66
AL556034.3 - 4 - 5 439 66
AY542307.1 - 4 - 5 395 66
AY687925.1 - 3 - 4 332 66
BC000384.2 - 1 379 2821006 66
BC006509.2 - 1 378 2961341 66
BC006520.2 - 1 378 2961341 66
BC007949.2 - 1 368 66
BC009865.2 - 1 383 3938904 66
BC013918.1 - 1 401 4048470 66
BC015813.1 - 1 383 3911362 66
BC018853.2 - 1 384 66
BC018857.2 - 1 383 66
BC019051.2 - 1 367 66
BC021974.2 - 1 131 66
BC024274.1 - 1 375 66
BC028179.1 - 1 395 5402023 66
BC031012.1 - 1 398 4654721 66
BC067738.1 - 1 397 5295336 66
BQ947806.1 - 4 - 5 523 66
BT006677.1 - 1 377 66
CR407625.1 - 1 377 66
M55409.1 - 1 - 2 142 66
X63526.1 - 4 - 5 396 66
Z11531.1 - 4 - 5 381 66
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BC007809.1 - 1 Chr. 7 - 133034515 - 133035924 [+] on Build GRCh38 66

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK300729.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EEF1G

Gene Ontology Categories:

Function(s) Process(es)

translational elongation
glutathione metabolic process
response to virus
cell-cell adhesion
translation elongation factor activity
glutathione transferase activity
protein binding
cadherin binding involved in cell-cell adhesion

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000524420.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000524420.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000524420.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000524420.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000524420.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000524420.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000524420.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000532986.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000532986.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000532986.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000532986.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000532986.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000532986.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 ENST00000532986.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 NM_001404.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 NM_001404.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 NM_001404.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 NM_001404.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 NM_001404.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 NM_001404.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 NM_001404.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 NM_001404.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 NM_001404.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 NM_001404.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 NM_001404.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 NM_001404.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 NM_001404.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 NM_001404.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 NM_001404.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 NM_001404.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 NM_001404.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 NR_037427.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013170.hg.1 NR_037427.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010805.hg.1 ENST00000421044.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008696.hg.1 ENST00000506566.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008986.hg.1 ENST00000457231.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008986.hg.1 ENST00000457231.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008986.hg.1 ENST00000457231.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012978.hg.1 ENST00000467631.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008201.hg.1 ENST00000434365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008201.hg.1 ENST00000434365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008201.hg.1 ENST00000434365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008201.hg.1 ENST00000434365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008201.hg.1 ENST00000434365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008201.hg.1 ENST00000434365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008201.hg.1 ENST00000580961.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008696.hg.1 ENST00000506566.1 Ensembl
Affymetrix Human Clariom D Assay TC0300010805.hg.1 ENST00000421044.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012978.hg.1 ENST00000467631.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000524420.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000524420.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000524420.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000524420.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000524420.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000524420.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000524420.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000532986.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000532986.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000532986.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000532986.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000532986.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000532986.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 ENST00000532986.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013170.hg.1 NM_001404.4 Refseq
Affymetrix Human Clariom D Assay TC1100013170.hg.1 NM_001404.4 Refseq
Affymetrix Human Clariom D Assay TC1100013170.hg.1 NM_001404.4 Refseq
Affymetrix Human Clariom D Assay TC1100013170.hg.1 NM_001404.4 Refseq
Affymetrix Human Clariom D Assay TC1100013170.hg.1 NM_001404.4 Refseq
Affymetrix Human Clariom D Assay TC1100013170.hg.1 NM_001404.4 Refseq
Affymetrix Human Clariom D Assay TC1100013170.hg.1 NM_001404.4 Refseq
Affymetrix Human Clariom D Assay TC1100013170.hg.1 NM_001404.4 Refseq
Affymetrix Human Clariom D Assay TC1100013170.hg.1 NM_001404.4 Refseq
Affymetrix Human Clariom D Assay TC1100013170.hg.1 NM_001404.4 Refseq
Affymetrix Human Clariom D Assay TC1100013170.hg.1 NM_001404.4 Refseq
Affymetrix Human Clariom D Assay TC1100013170.hg.1 NM_001404.4 Refseq
Affymetrix Human Clariom D Assay TC1100013170.hg.1 NM_001404.4 Refseq
Affymetrix Human Clariom D Assay TC1100013170.hg.1 NM_001404.4 Refseq
Affymetrix Human Clariom D Assay TC1100013170.hg.1 NM_001404.4 Refseq
Affymetrix Human Clariom D Assay TC1100013170.hg.1 NM_001404.4 Refseq
Affymetrix Human Clariom D Assay TC1100013170.hg.1 NM_001404.4 Refseq
Affymetrix Human Clariom D Assay TC1100013170.hg.1 NR_037427.1 Refseq
Affymetrix Human Clariom D Assay TC1100013170.hg.1 NR_037427.1 Refseq
Affymetrix Human Clariom D Assay TC0700008986.hg.1 ENST00000457231.1 Ensembl
Affymetrix Human Clariom D Assay TC0700008986.hg.1 ENST00000457231.1 Ensembl
Affymetrix Human Clariom D Assay TC0700008986.hg.1 ENST00000457231.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008201.hg.1 ENST00000434365.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008201.hg.1 ENST00000434365.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008201.hg.1 ENST00000434365.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008201.hg.1 ENST00000434365.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008201.hg.1 ENST00000434365.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008201.hg.1 ENST00000434365.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008201.hg.1 ENST00000580961.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0