This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02831111_g1
See other GRK6 GE Assays ›
Gene Symbol:
GRK6
Gene Name:
G protein-coupled receptor kinase 6
Gene Aliases:
GPRK6
Chromosome Location:
Chr.5: 177426496 - 177442849 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs02831111_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GRK6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001004105.2 NP_001004105.1 15 - 16 1835 79
GenBank mRNA AF040752.1 - 15 - 16 1678 79

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GRK6

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

desensitization of G-protein coupled receptor protein signaling pathway
protein phosphorylation
regulation of G-protein coupled receptor protein signaling pathway
negative regulation of anion channel activity
Wnt signaling pathway
G-protein coupled receptor kinase activity
protein binding
ATP binding
beta-adrenergic receptor kinase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline