This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02852647_m1
See other PHACTR3 GE Assays ›
Gene Symbol:
PHACTR3
Gene Name:
phosphatase and actin regulator 3
Gene Aliases:
C20orf101, H17739, PPP1R123, SCAPIN1, SCAPININ
Chromosome Location:
Chr.20: 59577497 - 59849829 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs02852647_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PHACTR3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001199505.1 NP_001186434.1 12 - 13 1662 111
NM_001199506.1 NP_001186435.1 12 - 13 1657 111
NM_001281507.1 NP_001268436.1 12 - 13 1761 111
NM_080672.4 NP_542403.1 12 - 13 2139 111
NM_183244.1 NP_899067.1 12 - 13 1782 111
NM_183246.1 NP_899069.1 11 - 12 1572 111
XM_011528525.2 XP_011526827.1 12 - 13 1768 111
XM_011528526.2 XP_011526828.1 11 - 12 1574 111
XM_017027626.1 XP_016883115.1 12 - 13 1871 111
XM_017027627.1 XP_016883116.1 12 - 13 1816 111
XM_017027628.1 XP_016883117.1 11 - 12 1464 111
XM_017027629.1 XP_016883118.1 11 - 12 1607 111
XM_017027630.1 XP_016883119.1 11 - 12 1435 111
XM_017027631.1 XP_016883120.1 12 - 13 1698 111
GenBank mRNA AB098521.1 - 12 - 13 1782 111
AB098522.1 - 11 - 12 1572 111
AJ311122.2 - 12 - 13 2138 111
AJ617581.1 - 12 - 13 1657 111
AK090812.1 - 1 - 2 1818 111
AK091632.1 - 12 - 13 1784 111
AK098788.1 - 12 - 13 1662 111
AK316047.1 - 12 - 13 1654 111
BC108303.1 - 12 - 13 2139 4179728 111
BC117362.1 - 12 - 13 2027 40125913 111
BC117364.1 - 12 - 13 2030 40125915 111

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PHACTR3

Panther Classification:

Molecular Function -

phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of catalytic activity
actin binding
protein phosphatase inhibitor activity
protein phosphatase 1 binding
protein phosphatase type 1 regulator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9mkk8:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0