This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02862720_m1
See other SSR3 GE Assays ›
Gene Symbol:
SSR3
Gene Name:
signal sequence receptor subunit 3
Gene Aliases:
TRAPG
Chromosome Location:
Chr.3: 156539553 - 156555200 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs02862720_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SSR3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001308204.1 NP_001295133.1 3 - 4 337 117
NM_001308205.1 NP_001295134.1 3 - 4 361 117
NM_007107.4 NP_009038.1 3 - 4 472 117
GenBank mRNA AF087907.1 - 3 - 4 420 117
AF110647.1 - 3 - 4 417 117
AK312935.1 - 3 - 4 379 117
BC017203.1 - 3 - 4 418 3618589 117
BT006669.1 - 3 - 4 361 117
DA283706.1 - 3 - 4 337 117
DA725890.1 - 3 - 4 363 117
DA750309.1 - 3 - 4 456 117

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SSR3

Gene Ontology Categories:

Function(s)

SRP-dependent cotranslational protein targeting to membrane

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4krp5:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0