This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02873161_m1
See other PPP1R14D GE Assays ›
Gene Symbol:
PPP1R14D
Gene Name:
protein phosphatase 1 regulatory inhibitor subunit 14D
Gene Aliases:
CPI17-like, GBPI-1, GBPI1
Chromosome Location:
Chr.15: 40815445 - 40828793 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs02873161_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPP1R14D
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001130143.1 NP_001123615.1 4 - 5 556 79
NM_017726.7 NP_060196.1 3 - 4 440 79
GenBank mRNA AI963626.1 - 1 - 2 325 79
AK000258.1 - 3 - 4 439 79
AY050671.1 - 3 - 4 468 79
BC096716.1 - 3 - 4 446 40003029 79
BC098124.1 - 3 - 4 446 40003027 79
BC098150.1 - 4 - 5 562 79
BC098341.1 - 4 - 5 562 79

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PPP1R14D

Panther Classification:

Molecular Function -

phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

regulation of protein dephosphorylation
regulation of phosphorylation
negative regulation of catalytic activity
protein serine/threonine phosphatase inhibitor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0