This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02883646_gH
See other HYI GE Assays ›
Gene Symbol:
HYI
Gene Name:
hydroxypyruvate isomerase (putative)
Gene Aliases:
HT036
Chromosome Location:
Chr.1: 43450607 - 43454241 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs02883646_gH
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HYI
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001190880.2 NP_001177809.1 6 - 7 820 70
NM_001243526.1 NP_001230455.1 7 - 8 895 70
NM_031207.5 NP_112484.3 6 - 7 820 70
XM_005271239.4 XP_005271296.1 7 - 8 1147 70
XM_005271240.4 XP_005271297.1 6 - 7 1045 70
XM_006710937.3 XP_006711000.1 7 - 8 1147 70
XM_011542222.2 XP_011540524.1 6 - 7 1045 70
GenBank mRNA AF284750.1 - 6 - 7 695 70
AF284751.1 - 6 - 7 695 70
AK096244.1 - 4 - 5 1896 70
AY037165.1 - 6 - 7 794 70
BC006140.1 - 6 - 7 506 3928690 70
BC019041.2 - 6 - 7 622 4586546 70
CA433653.1 - 1 - 2 412 70
CD672533.1 - 2 - 3 312 70

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene HYI

Panther Classification:

Molecular Function -

isomerase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
protein binding
hydroxypyruvate isomerase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0