This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02888324_m1
See other FGF14 GE Assays ›
Gene Symbol:
FGF14
Gene Name:
fibroblast growth factor 14
Gene Aliases:
FGF-14, FHF-4, FHF4, SCA27
Chromosome Location:
Chr.13: 101720855 - 102402428 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs02888324_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FGF14
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001321937.1 NP_001308866.1 1 - 2 562 98
NM_001321945.1 NP_001308874.1 2 - 3 434 98
NM_001321947.1 NP_001308876.1 2 - 3 358 98
NM_001321948.1 NP_001308877.1 2 - 3 430 98
NM_175929.2 NP_787125.1 1 - 2 304 98
GenBank mRNA AY188178.1 - 1 - 2 304 98
BC100920.1 - 1 - 2 303 40003901 98
BC100921.1 - 1 - 2 303 40003902 98
BC100922.2 - 1 - 2 303 40003903 98

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001321947.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene FGF14

Panther Classification:

Molecular Function -

growth factor intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of defense response to virus by host
signal transduction
cell-cell signaling
nervous system development
regulation of synaptic plasticity
regulation of postsynaptic membrane potential
xenophagy
positive regulation of high voltage-gated calcium channel activity
regulation of synaptic vesicle recycling
protein binding
growth factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline