This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02888397_m1
See other FTSJ1 GE Assays ›
Gene Symbol:
FTSJ1
Gene Name:
FtsJ RNA methyltransferase homolog 1 (E. coli)
Gene Aliases:
CDLIV, JM23, MRX44, MRX9, SPB1, TRMT7
Chromosome Location:
Chr.X: 48476021 - 48486364 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs02888397_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FTSJ1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177439.2 NP_803188.1 10 - 11 1216 70
XM_005272595.1 XP_005272652.1 9 - 10 1065 70
XM_011543895.1 XP_011542197.1 9 - 10 1065 70
XM_017029382.1 XP_016884871.1 10 - 11 1061 70
GenBank mRNA AF063015.1 - 8 - 9 703 70
AK024023.1 - 10 - 11 1076 70

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene FTSJ1

Panther Classification:

Molecular Function -

RNA metabolism protein RNA methyltransferase nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

tRNA nucleoside ribose methylation
cytoplasmic translation
tRNA methylation
tRNA methyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-m2kmk:80/100.66.77.245:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0