This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03037374_m1
See other NPC1L1 GE Assays ›
Gene Symbol:
NPC1L1
Gene Name:
NPC1 like intracellular cholesterol transporter 1
Gene Aliases:
NPC11L1
Chromosome Location:
Chr.7: 44512440 - 44541650 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03037374_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NPC1L1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001101648.1 NP_001095118.1 2 - 3 1636 80
NM_001300967.1 NP_001287896.1 2 - 3 1636 80
NM_013389.2 NP_037521.2 2 - 3 1636 80
XM_011515326.2 XP_011513628.1 2 - 3 1682 80
XM_011515327.2 XP_011513629.1 2 - 3 1971 80
GenBank mRNA AF192522.1 - 2 - 3 1636 80
AF192523.1 - 2 - 3 1636 80
AY437865.1 - 2 - 3 1580 80
AY515256.1 - 2 - 3 1580 80
BC117178.1 - 2 - 3 1634 40125729 80
BC143756.1 - 2 - 3 1634 9052270 80
FJ481111.1 - 2 - 3 1580 80

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NPC1L1

Gene Ontology Categories:

Function(s) Process(es)

cholesterol biosynthetic process
sterol transport
intestinal cholesterol absorption
cholesterol transport
lipoprotein metabolic process
response to drug
protein binding
drug binding
Rab GTPase binding
myosin V binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0