This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03054896_m1
See other CEACAM20 GE Assays ›
Gene Symbol:
CEACAM20
Gene Name:
carcinoembryonic antigen related cell adhesion molecule 20
Gene Aliases:
UNQ9366
Chromosome Location:
Chr.19: 44503838 - 44530100 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03054896_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CEACAM20
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001102597.2 NP_001096067.2 6 - 7 1326 107
NM_001102600.2 NP_001096070.2 6 - 7 1326 107
XM_011526429.2 XP_011524731.1 7 - 8 1980 107
XM_011526430.2 XP_011524732.1 7 - 8 1981 107
XM_011526431.2 XP_011524733.1 7 - 8 1982 107
XM_011526432.2 XP_011524734.1 3 - 4 644 107
GenBank mRNA AY358129.1 - 6 - 7 1589 107
EU008600.1 - 6 - 7 1326 107
EU008603.1 - 6 - 7 1326 107

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 G-protein coupled receptor Homo sapiens Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-6dxlp:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0