This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03063375_ft
See other TMPRSS2-ERG FUSION GE Assays ›
Gene Symbol:
TMPRSS2-ERG fusion
Gene Name:
FUSION
Gene Aliases:
ERG, TMPRSS2
Chromosome Location:
-
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03063375_ft
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMPRSS2-ERG fusion
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA DQ204772.1 - 49 106

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Fusion Homo sapiens Inventoried

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline