This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03297855_m1
See other ST7-AS2 GE Assays ›
Gene Symbol:
ST7-AS2
Gene Name:
ST7 antisense RNA 2
Gene Aliases:
ST7AS2, ST7OT2
Chromosome Location:
Chr.7: 117072072 - 117145592 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03297855_m1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 360.22
360.22
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ST7-AS2
Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_002331.3
3 - 4 436 88
GenBank mRNA AF400045.1 - LIT1899
ST7OT2
3 - 4 426 88
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AA868610.1 - 3 - 4 Chr. 7 - 117112292 - 117130598 [+] on Build GRCh38 88
GenBankmRNA BX097853.1 - 3 - 4 403 Chr. 7 - 117112518 - 117145559 [-] on Build GRCh38 88

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 6132 MIR6132 hsa-mir-6132 -108.0kb Anti-sense Chr. 7 - 117020211 - 117020319 on NCBI Build GRCh38
Downstream ST7 overlapping transcript 3 ST7-OT3 NCRNA00026,
ST7,
ST7OT3
52.0kb Anti-sense Chr. 7 - 117182681 - 117209937 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700012363.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07001792.hg.1

Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000433969.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000433969.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000433969.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000433969.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000442719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000442719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000442719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000442719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000442719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000442719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000446784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000446784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000446784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000446784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000446784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000446784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000446784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000446784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_002331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_002331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_002331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_002331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_002331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_002331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_002331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_109980.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_109980.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_109980.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_109980.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_109980.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_109981.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_109981.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_109981.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_109981.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_109981.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_109981.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07027823.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07027823.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07027823.hg.1 ENST00000442719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07027823.hg.1 NR_002331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07027823.hg.1 TC07001792.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR07027819.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07027819.hg.1 ENST00000442719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07027819.hg.1 NR_002331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07027819.hg.1 TC07001792.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR07027821.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07027821.hg.1 NR_002331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07027821.hg.1 TC07001792.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR07027824.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07027824.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07027824.hg.1 ENST00000442719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07027824.hg.1 NR_002331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07027824.hg.1 TC07001792.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR07027822.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07027822.hg.1 NR_002331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07027822.hg.1 TC07001792.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000433969.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000433969.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000433969.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000433969.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000442719.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000442719.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000442719.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000442719.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000442719.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000442719.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000446784.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000446784.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000446784.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000446784.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000446784.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000446784.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000446784.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000446784.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_002331.3 Refseq
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_002331.3 Refseq
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_002331.3 Refseq
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_002331.3 Refseq
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_002331.3 Refseq
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_002331.3 Refseq
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_002331.3 Refseq
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_109980.1 Refseq
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_109980.1 Refseq
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_109980.1 Refseq
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_109980.1 Refseq
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_109980.1 Refseq
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_109981.1 Refseq
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_109981.1 Refseq
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_109981.1 Refseq
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_109981.1 Refseq
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_109981.1 Refseq
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_109981.1 Refseq
Affymetrix Human Clariom D Assay PSR0700189069.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700189069.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700189069.hg.1 NR_002331.3 Refseq
Affymetrix Human Clariom D Assay PSR0700189069.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700189069.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700189069.hg.1 NR_002331.3 Refseq
Affymetrix Human Clariom D Assay PSR0700189069.hg.1 TC0700012363.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0700189068.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700189068.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700189068.hg.1 NR_002331.3 Refseq
Affymetrix Human Clariom D Assay PSR0700189068.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700189068.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700189068.hg.1 NR_002331.3 Refseq
Affymetrix Human Clariom D Assay PSR0700189068.hg.1 TC0700012363.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-hrdnk:80/100.66.78.145:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0