This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03309334_s1
See other PER4 GE Assays ›
Gene Symbol:
PER4
Gene Name:
period circadian clock 3 pseudogene
Gene Aliases:
-
Chromosome Location:
Chr.7: 9634270 - 9635817 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03309334_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PER4
Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_002790.3
1 1147 111
GenBank mRNA AF348410.1 - LIT2066
Per4 pseudogene
1 1148 111

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream neurexophilin 1 NXPH1 NPH1,
Nbla00697
-881.3kb Sense Chr. 7 - 8433955 - 8752963 on NCBI Build GRCh38
Downstream uncharacterized LOC100131472 LOC100131472 1.1mb Sense Chr. 7 - 10702450 - 10707538 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700006699.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07000084.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0700006830.hg.1 ENST00000412563.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006830.hg.1 ENST00000412563.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006830.hg.1 ENST00000412563.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006830.hg.1 ENST00000412563.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006830.hg.1 ENST00000412563.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006830.hg.1 ENST00000412563.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006830.hg.1 ENST00000412563.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006699.hg.1 ENST00000605169.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006699.hg.1 ENST00000605169.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006699.hg.1 NR_002790.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006699.hg.1 NR_002790.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006699.hg.1 NR_002790.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006699.hg.1 NR_002790.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07001416.hg.1 NR_002790.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07001416.hg.1 TC07000084.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0700006699.hg.1 ENST00000605169.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006699.hg.1 ENST00000605169.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006699.hg.1 NR_002790.3 Refseq
Affymetrix Human Clariom D Assay TC0700006699.hg.1 NR_002790.3 Refseq
Affymetrix Human Clariom D Assay TC0700006699.hg.1 NR_002790.3 Refseq
Affymetrix Human Clariom D Assay TC0700006699.hg.1 NR_002790.3 Refseq
Affymetrix Human Clariom D Assay TC0700006830.hg.1 ENST00000412563.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006830.hg.1 ENST00000412563.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006830.hg.1 ENST00000412563.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006830.hg.1 ENST00000412563.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006830.hg.1 ENST00000412563.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006830.hg.1 ENST00000412563.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006830.hg.1 ENST00000412563.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700146331.hg.1 ENST00000605169.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700146331.hg.1 NR_002790.3 Refseq
Affymetrix Human Clariom D Assay PSR0700146331.hg.1 ENST00000605169.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700146331.hg.1 NR_002790.3 Refseq
Affymetrix Human Clariom D Assay PSR0700146331.hg.1 TC0700006699.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0700146329.hg.1 ENST00000605169.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700146329.hg.1 NR_002790.3 Refseq
Affymetrix Human Clariom D Assay PSR0700146329.hg.1 ENST00000605169.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700146329.hg.1 NR_002790.3 Refseq
Affymetrix Human Clariom D Assay PSR0700146329.hg.1 TC0700006699.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0700146330.hg.1 NR_002790.3 Refseq
Affymetrix Human Clariom D Assay PSR0700146330.hg.1 NR_002790.3 Refseq
Affymetrix Human Clariom D Assay PSR0700146330.hg.1 TC0700006699.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline