This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03453838_m1
See other ST7-AS2 GE Assays ›
Gene Symbol:
ST7-AS2
Gene Name:
ST7 antisense RNA 2
Gene Aliases:
ST7AS2, ST7OT2
Chromosome Location:
Chr.7: 117072072 - 117145592 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03453838_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ST7-AS2
Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_109980.1
2 - 3 259 133
GenBank mRNA AF400040.1 - LIT1999
ST7OT2
3 - 4 449 133
AF400042.1 - LIT2001
ST7OT2
2 - 3 280 133
CB054673.1 -
2 - 3 268 133
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BI262734.1 - 2 - 3 280 Chr. 7 - 117072070 - 117090781 [-] on Build GRCh38 133
GenBankmRNA BI262732.1 - 3 - 4 449 Chr. 7 - 117072424 - 117145558 [-] on Build GRCh38 133

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 6132 MIR6132 hsa-mir-6132 -53.5kb Anti-sense Chr. 7 - 117020211 - 117020319 on NCBI Build GRCh38
Downstream ST7 overlapping transcript 3 ST7-OT3 NCRNA00026,
ST7,
ST7OT3
101.6kb Anti-sense Chr. 7 - 117182681 - 117209937 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0