This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03453986_s1
See other DSTNP2 GE Assays ›
Gene Symbol:
DSTNP2
Gene Name:
destrin, actin depolymerizing factor pseudogene 2
Gene Aliases:
-
Chromosome Location:
Chr.12: 6884682 - 6885786 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03453986_s1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer:
434.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DSTNP2
Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_033796.2
1 150 91
GenBank mRNA U72518.1 - LIT2102
destrin-2 pseudogene
1 150 91
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CN483017.1 - 1 227 Chr. 12 - 6884605 - 6885242 [+] on Build GRCh38 91

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream uncharacterized LOC105369632 LOC105369632 Anti-sense Chr. 12 - 6875764 - 6889358 on NCBI Build GRCh38
Downstream leucine rich repeat containing 23 LRRC23 LRPB7 19.0kb Sense Chr. 12 - 6904733 - 6914242 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200012589.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12000098.hg.1

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ArrayStar ABI

Array Name Probe IDs Targets Target Type
Agilent Human Whole Genome ChIP Tiling Array A_17_P16527553 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P28606976 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P08178062 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P08178046 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P28606967 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P28606959 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P28606983 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P16527551 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P16527549 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P28606971 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P08178055 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P08178052 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P08178060 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P28606968 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P28606970 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P08178053 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P28606977 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P08178045 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3279379 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 55566 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3279379 - -
SurePrint G3 Human GE Microarray G4851A A_33_P3279379 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02565492 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.0 ST Array 7953519 - -
Affymetrix Human Gene 1.1 ST Array 7953518 - -
Affymetrix HT HG-U133+ PM Array Plate 211325_PM_x_at - -
Affymetrix Human Genome U219 Array 11760438_at - -
Affymetrix Human Genome U219 Array 11760439_PM_x_at - -
Affymetrix Human Genome U219 Array 11760439_x_at - -
Affymetrix Human Genome U219 Array 11760438_PM_at - -
Affymetrix Human Transcriptome Array 2.0 PSR12001254.hg.1 NR_033796.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12001254.hg.1 TC12000098.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 ENST00000274820.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 ENST00000274820.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 ENST00000412023.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 ENST00000412023.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 ENST00000412023.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 ENST00000412023.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 ENST00000412023.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 ENST00000412023.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 ENST00000412023.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 ENST00000412023.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 ENST00000412023.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 ENST00000421824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 ENST00000421824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 ENST00000421824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 ENST00000421824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 ENST00000421824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 ENST00000421824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 ENST00000421824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 ENST00000441655.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 ENST00000441655.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 ENST00000441655.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 ENST00000451612.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 ENST00000451612.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 ENST00000451612.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 ENST00000451612.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 ENST00000504533.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 ENST00000504533.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 ENST00000602547.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 ENST00000602547.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 ENST00000602547.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 ENST00000602547.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 ENST00000602547.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 ENST00000602547.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 NR_002803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 NR_002803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 NR_002803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 NR_033796.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 NR_033796.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 NR_033796.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 NR_033796.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 NR_033796.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012589.hg.1 NR_033796.2 Refseq
Affymetrix Human Clariom D Assay PSR1200199002.hg.1 ENST00000602547.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200199002.hg.1 NR_033796.2 Refseq
Affymetrix Human Clariom D Assay PSR1200199002.hg.1 ENST00000602547.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200199002.hg.1 NR_033796.2 Refseq
Affymetrix Human Clariom D Assay PSR1200199002.hg.1 TC1200012589.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200199006.hg.1 ENST00000602547.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200199006.hg.1 NR_033796.2 Refseq
Affymetrix Human Clariom D Assay PSR1200199006.hg.1 ENST00000602547.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200199006.hg.1 NR_033796.2 Refseq
Affymetrix Human Clariom D Assay PSR1200199006.hg.1 TC1200012589.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200199003.hg.1 ENST00000504533.2 Ensembl
Affymetrix Human Clariom D Assay PSR1200199003.hg.1 ENST00000602547.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200199003.hg.1 NR_033796.2 Refseq
Affymetrix Human Clariom D Assay PSR1200199003.hg.1 ENST00000504533.2 Ensembl
Affymetrix Human Clariom D Assay PSR1200199003.hg.1 ENST00000602547.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200199003.hg.1 NR_033796.2 Refseq
Affymetrix Human Clariom D Assay PSR1200199003.hg.1 TC1200012589.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200199005.hg.1 ENST00000504533.2 Ensembl
Affymetrix Human Clariom D Assay PSR1200199005.hg.1 ENST00000602547.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200199005.hg.1 NR_033796.2 Refseq
Affymetrix Human Clariom D Assay PSR1200199005.hg.1 ENST00000504533.2 Ensembl
Affymetrix Human Clariom D Assay PSR1200199005.hg.1 ENST00000602547.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200199005.hg.1 NR_033796.2 Refseq
Affymetrix Human Clariom D Assay PSR1200199005.hg.1 TC1200012589.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1200012589.hg.1 ENST00000274820.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012589.hg.1 ENST00000274820.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012589.hg.1 ENST00000412023.5 Ensembl
Affymetrix Human Clariom D Assay TC1200012589.hg.1 ENST00000412023.5 Ensembl
Affymetrix Human Clariom D Assay TC1200012589.hg.1 ENST00000412023.5 Ensembl
Affymetrix Human Clariom D Assay TC1200012589.hg.1 ENST00000412023.5 Ensembl
Affymetrix Human Clariom D Assay TC1200012589.hg.1 ENST00000412023.5 Ensembl
Affymetrix Human Clariom D Assay TC1200012589.hg.1 ENST00000412023.5 Ensembl
Affymetrix Human Clariom D Assay TC1200012589.hg.1 ENST00000412023.5 Ensembl
Affymetrix Human Clariom D Assay TC1200012589.hg.1 ENST00000412023.5 Ensembl
Affymetrix Human Clariom D Assay TC1200012589.hg.1 ENST00000412023.5 Ensembl
Affymetrix Human Clariom D Assay TC1200012589.hg.1 ENST00000421824.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012589.hg.1 ENST00000421824.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012589.hg.1 ENST00000421824.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012589.hg.1 ENST00000421824.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012589.hg.1 ENST00000421824.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012589.hg.1 ENST00000421824.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012589.hg.1 ENST00000421824.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012589.hg.1 ENST00000441655.2 Ensembl
Affymetrix Human Clariom D Assay TC1200012589.hg.1 ENST00000441655.2 Ensembl
Affymetrix Human Clariom D Assay TC1200012589.hg.1 ENST00000441655.2 Ensembl
Affymetrix Human Clariom D Assay TC1200012589.hg.1 ENST00000451612.5 Ensembl
Affymetrix Human Clariom D Assay TC1200012589.hg.1 ENST00000451612.5 Ensembl
Affymetrix Human Clariom D Assay TC1200012589.hg.1 ENST00000451612.5 Ensembl
Affymetrix Human Clariom D Assay TC1200012589.hg.1 ENST00000451612.5 Ensembl
Affymetrix Human Clariom D Assay TC1200012589.hg.1 ENST00000504533.2 Ensembl
Affymetrix Human Clariom D Assay TC1200012589.hg.1 ENST00000504533.2 Ensembl
Affymetrix Human Clariom D Assay TC1200012589.hg.1 ENST00000602547.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012589.hg.1 ENST00000602547.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012589.hg.1 ENST00000602547.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012589.hg.1 ENST00000602547.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012589.hg.1 ENST00000602547.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012589.hg.1 ENST00000602547.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012589.hg.1 NR_002803.2 Refseq
Affymetrix Human Clariom D Assay TC1200012589.hg.1 NR_002803.2 Refseq
Affymetrix Human Clariom D Assay TC1200012589.hg.1 NR_002803.2 Refseq
Affymetrix Human Clariom D Assay TC1200012589.hg.1 NR_033796.2 Refseq
Affymetrix Human Clariom D Assay TC1200012589.hg.1 NR_033796.2 Refseq
Affymetrix Human Clariom D Assay TC1200012589.hg.1 NR_033796.2 Refseq
Affymetrix Human Clariom D Assay TC1200012589.hg.1 NR_033796.2 Refseq
Affymetrix Human Clariom D Assay TC1200012589.hg.1 NR_033796.2 Refseq
Affymetrix Human Clariom D Assay TC1200012589.hg.1 NR_033796.2 Refseq
Affymetrix Human Clariom D Assay PSR1200199001.hg.1 ENST00000421824.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200199001.hg.1 ENST00000602547.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200199001.hg.1 NR_033796.2 Refseq
Affymetrix Human Clariom D Assay PSR1200199001.hg.1 ENST00000421824.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200199001.hg.1 ENST00000602547.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200199001.hg.1 NR_033796.2 Refseq
Affymetrix Human Clariom D Assay PSR1200199001.hg.1 TC1200012589.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200199004.hg.1 ENST00000602547.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200199004.hg.1 NR_033796.2 Refseq
Affymetrix Human Clariom D Assay PSR1200199004.hg.1 ENST00000602547.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200199004.hg.1 NR_033796.2 Refseq
Affymetrix Human Clariom D Assay PSR1200199004.hg.1 TC1200012589.hg.1 Clariom D TC
Human Genome U133 Plus 2.0 Array 211325_PM_x_at - -
Human Genome U133 Plus 2.0 Array 211325_x_at - -
Human Genome U133A 2.0 Array 211325_PM_x_at - -
Human Genome U133A 2.0 Array 211325_x_at - -
AffymetrixPathwork Diagnostics Pathchip pw594017 - -
Human Genome U95 Set 31605_PM_at - -
Human Genome U95 Set 31605_at - -
Affymetrix Human Genome U133 Plus 2.0 Array 211325_x_at1 - -
Affymetrix Human Genome U133 Plus 2.0 Array 211325_x_at8 - -
Affymetrix Human Genome U133 Plus 2.0 Array 211325_x_at3 - -
Affymetrix Human Genome U133 Plus 2.0 Array 211325_x_at6 - -
Affymetrix Human Genome U133 Plus 2.0 Array 211325_x_at7 - -
Affymetrix Human Genome U133 Plus 2.0 Array 211325_x_at11 - -
Affymetrix Human Genome U133 Plus 2.0 Array 211325_x_at2 - -
Affymetrix Human Genome U133 Plus 2.0 Array 211325_x_at10 - -
Affymetrix Human Genome U133 Plus 2.0 Array 211325_x_at4 - -
Affymetrix Human Genome U133 Plus 2.0 Array 211325_x_at5 - -
Affymetrix Human Genome U133 Plus 2.0 Array 211325_x_at9 - -
Human Genome U95 Set 63906_PM_f_at - -
Human Genome U95 Set 63705_f_at - -
Human Genome U95 Set 63705_PM_f_at - -
Human Genome U95 Set 63906_f_at - -
Human Genome U133 Set 211325_x_at - -
Human Genome U133 Set 211325_PM_x_at - -
Array Name Probe IDs Targets Target Type
Arraystar Human LncRNA microarray V2.0 ASHG19A3A048292 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A048292 - -
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 184839 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline