Assay Details
| Gene Symbol: TERC | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_001566.1 | LIT2117 telomerase RNA |
1 | 54 | 100 | ||
| GenBank mRNA | BC128029.1 | - | 1 | 54 | 100 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001982.hg.1
Set Membership: |
|

TERC
Genomic Map