Assay Details
| Gene Symbol: PBDC1 | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY927616.1 | - | LIT3604 AY927616 |
1 - 2 | 239 | 103 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MAGE family member E2 | MAGEE2 | HCA3 | -387.7kb | Anti-sense | Chr. X - 75782988 - 75785244 on NCBI Build GRCh38 |
| Downstream | MAGE family member E1 | MAGEE1 |
DAMAGE, HCA1 |
254.0kb | Sense | Chr. X - 76427653 - 76431353 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | ENST00000373357.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | ENST00000373357.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | ENST00000373357.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | ENST00000373357.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | ENST00000373357.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | ENST00000373357.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | ENST00000373357.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | ENST00000373357.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | ENST00000373357.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | ENST00000373358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | ENST00000373358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | ENST00000373358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | ENST00000373358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | ENST00000373358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | ENST00000373358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | ENST00000373358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | ENST00000373358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | ENST00000373358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | ENST00000373358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | ENST00000373358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | NM_001300888.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | NM_001300888.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | NM_001300888.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | NM_001300888.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | NM_001300888.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | NM_001300888.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | NM_001300888.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | NM_001300888.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | NM_001300888.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | NM_001300888.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | NM_001300888.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | NM_016500.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | NM_016500.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | NM_016500.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | NM_016500.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | NM_016500.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | NM_016500.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | NM_016500.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | NM_016500.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | NM_016500.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | NM_016500.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007686.hg.1 | NM_016500.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | ENST00000373357.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | ENST00000373357.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | ENST00000373357.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | ENST00000373357.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | ENST00000373357.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | ENST00000373357.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | ENST00000373357.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | ENST00000373357.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | ENST00000373357.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | ENST00000373358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | ENST00000373358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | ENST00000373358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | ENST00000373358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | ENST00000373358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | ENST00000373358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | ENST00000373358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | ENST00000373358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | ENST00000373358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | ENST00000373358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | ENST00000373358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | NM_001300888.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | NM_001300888.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | NM_001300888.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | NM_001300888.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | NM_001300888.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | NM_001300888.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | NM_001300888.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | NM_001300888.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | NM_001300888.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | NM_001300888.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | NM_001300888.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | NM_016500.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | NM_016500.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | NM_016500.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | NM_016500.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | NM_016500.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | NM_016500.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | NM_016500.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | NM_016500.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | NM_016500.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | NM_016500.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007686.hg.1 | NM_016500.3 | Refseq |

PBDC1
Genomic Map