This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03461947_mH
See other PMCHL1 GE Assays ›
Gene Symbol:
PMCHL1
Gene Name:
pro-melanin concentrating hormone like 1 (pseudogene)
Gene Aliases:
-
Chromosome Location:
Chr.5: 22142352 - 22152272 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03461947_mH
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PMCHL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_003921.1 2 - 3 310 130
GenBank mRNA AY008409.1 - 2 - 3 310 130

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream glucuronidase,
beta pseudogene 1
GUSBP1 -554.7kb Sense Chr. 5 - 21459480 - 21589372 on NCBI Build GRCh38
Downstream PR domain 9 PRDM9 MEISETZ,
MSBP3,
PFM6,
ZNF899
1.4mb Sense Chr. 5 - 23507609 - 23528597 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500006935.hg.1
This assay targets the same location as HTA transcript cluster(s) TC05000111.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

peptide hormone intercellular signal molecule

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0500006935.hg.1 ENST00000418902.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006935.hg.1 ENST00000418902.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006935.hg.1 ENST00000418902.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006935.hg.1 ENST00000418902.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006935.hg.1 ENST00000418902.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006935.hg.1 ENST00000418902.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006935.hg.1 ENST00000418902.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006935.hg.1 ENST00000418902.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006935.hg.1 ENST00000423102.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006935.hg.1 ENST00000423102.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006935.hg.1 ENST00000423301.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006935.hg.1 ENST00000423301.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006935.hg.1 ENST00000423301.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006935.hg.1 ENST00000423301.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006935.hg.1 ENST00000423301.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006935.hg.1 ENST00000423301.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006935.hg.1 ENST00000430902.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006935.hg.1 ENST00000430902.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006935.hg.1 ENST00000430902.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006935.hg.1 ENST00000430902.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006935.hg.1 ENST00000430902.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006935.hg.1 NR_003921.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006935.hg.1 NR_003921.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006935.hg.1 NR_003921.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006935.hg.1 NR_003921.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05001188.hg.1 ENST00000418902.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05001188.hg.1 NR_003921.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05001188.hg.1 TC05000111.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0500006935.hg.1 ENST00000418902.5 Ensembl
Affymetrix Human Clariom D Assay TC0500006935.hg.1 ENST00000418902.5 Ensembl
Affymetrix Human Clariom D Assay TC0500006935.hg.1 ENST00000418902.5 Ensembl
Affymetrix Human Clariom D Assay TC0500006935.hg.1 ENST00000418902.5 Ensembl
Affymetrix Human Clariom D Assay TC0500006935.hg.1 ENST00000418902.5 Ensembl
Affymetrix Human Clariom D Assay TC0500006935.hg.1 ENST00000418902.5 Ensembl
Affymetrix Human Clariom D Assay TC0500006935.hg.1 ENST00000418902.5 Ensembl
Affymetrix Human Clariom D Assay TC0500006935.hg.1 ENST00000418902.5 Ensembl
Affymetrix Human Clariom D Assay TC0500006935.hg.1 ENST00000423102.2 Ensembl
Affymetrix Human Clariom D Assay TC0500006935.hg.1 ENST00000423102.2 Ensembl
Affymetrix Human Clariom D Assay TC0500006935.hg.1 ENST00000423301.5 Ensembl
Affymetrix Human Clariom D Assay TC0500006935.hg.1 ENST00000423301.5 Ensembl
Affymetrix Human Clariom D Assay TC0500006935.hg.1 ENST00000423301.5 Ensembl
Affymetrix Human Clariom D Assay TC0500006935.hg.1 ENST00000423301.5 Ensembl
Affymetrix Human Clariom D Assay TC0500006935.hg.1 ENST00000423301.5 Ensembl
Affymetrix Human Clariom D Assay TC0500006935.hg.1 ENST00000423301.5 Ensembl
Affymetrix Human Clariom D Assay TC0500006935.hg.1 ENST00000430902.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006935.hg.1 ENST00000430902.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006935.hg.1 ENST00000430902.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006935.hg.1 ENST00000430902.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006935.hg.1 ENST00000430902.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006935.hg.1 NR_003921.1 Refseq
Affymetrix Human Clariom D Assay TC0500006935.hg.1 NR_003921.1 Refseq
Affymetrix Human Clariom D Assay TC0500006935.hg.1 NR_003921.1 Refseq
Affymetrix Human Clariom D Assay TC0500006935.hg.1 NR_003921.1 Refseq
Affymetrix Human Clariom D Assay PSR0500146996.hg.1 ENST00000418902.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500146996.hg.1 ENST00000423301.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500146996.hg.1 ENST00000430902.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500146996.hg.1 NR_003921.1 Refseq
Affymetrix Human Clariom D Assay PSR0500146996.hg.1 ENST00000418902.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500146996.hg.1 ENST00000423301.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500146996.hg.1 ENST00000430902.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500146996.hg.1 NR_003921.1 Refseq
Affymetrix Human Clariom D Assay PSR0500146996.hg.1 TC0500006935.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline