This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03463829_s1
See other ZNF137P GE Assays ›
Gene Symbol:
ZNF137P
Gene Name:
zinc finger protein 137, pseudogene
Gene Aliases:
ZNF137, pHZ-30
Chromosome Location:
Chr.19: 52596684 - 52600152 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03463829_s1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 407.65
407.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZNF137P
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_023311.1 3 2394 150
GenBank mRNA BM988340.1 - 1 150
U09414.1 - 3 2428 150
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AA766074.1 - 1 Chr. 19 - 52599812 - 52600149 [-] on Build GRCh38 150
GenBankmRNA AA503545.1 - 1 Chr. 19 - 52599641 - 52600148 [-] on Build GRCh38 150
GenBankmRNA AA043458.1 - 1 Chr. 19 - 52599708 - 52600149 [-] on Build GRCh38 150
GenBankmRNA BX103685.1 - 1 501 Chr. 19 - 52599562 - 52600140 [+] on Build GRCh38 150
GenBankmRNA BP343181.1 - 286 Chr. - - [-] on Build 150
GenBankmRNA AA765442.1 - 1 Chr. 19 - 52599936 - 52600149 [-] on Build GRCh38 150

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream zinc finger protein 701 ZNF701 -12.0kb Sense Chr. 19 - 52570273 - 52627269 on NCBI Build GRCh38
Downstream zinc finger protein 83 ZNF83 HPF1,
ZNF816B
12.2kb Anti-sense Chr. 19 - 52612364 - 52690581 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1900011794.hg.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

MicroArray Probes:

Agilent Affymetrix ArrayStar ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 394951 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 405635 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 363083 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 139318 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 244679 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3222753 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_23_P208238 - -
Agilent Whole Human Genome Oligo Microarray 4x44K 39377 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 7497 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 18025 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3222753 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_23_P208238 - -
SurePrint G3 Human GE Microarray G4851A A_23_P208238 - -
SurePrint G3 Human GE Microarray G4851A A_33_P3222753 - -
Agilent Whole Chr array_18 19 21 A_17_P10996284 - -
Agilent Whole Chr array_18 19 21 A_17_P10996264 - -
Agilent Whole Chr array_18 19 21 A_17_P10996269 - -
Agilent Whole Chr array_18 19 21 A_17_P31635918 - -
Agilent Whole Chr array_18 19 21 A_17_P10996273 - -
Agilent Whole Chr array_18 19 21 A_17_P10996290 - -
Agilent Whole Chr array_18 19 21 A_17_P10996289 - -
Agilent Whole Chr array_18 19 21 A_17_P10996301 - -
Agilent Whole Chr array_18 19 21 A_17_P17156717 - -
Agilent Whole Chr array_18 19 21 A_17_P10996297 - -
Agilent Whole Chr array_18 19 21 A_17_P31635861 - -
Agilent Whole Chr array_18 19 21 A_17_P31635862 - -
Agilent Whole Chr array_18 19 21 A_17_P10996280 - -
Agilent Whole Chr array_18 19 21 A_17_P31635868 - -
Agilent Whole Chr array_18 19 21 A_17_P31635874 - -
Agilent Whole Chr array_18 19 21 A_17_P10996292 - -
Agilent Whole Chr array_18 19 21 A_17_P31635910 - -
Agilent Whole Chr array_18 19 21 A_17_P31635890 - -
Agilent Whole Chr array_18 19 21 A_17_P10996299 - -
Agilent Whole Chr array_18 19 21 A_17_P31635884 - -
Agilent Whole Chr array_18 19 21 A_17_P17156687 - -
Agilent Whole Chr array_18 19 21 A_17_P10996272 - -
Agilent Whole Chr array_18 19 21 A_17_P31635907 - -
Agilent Whole Chr array_18 19 21 A_17_P31635864 - -
Agilent Whole Chr array_18 19 21 A_17_P10996285 - -
Agilent Whole Chr array_18 19 21 A_17_P10996276 - -
Agilent Whole Chr array_18 19 21 A_17_P31635905 - -
Agilent Whole Chr array_18 19 21 A_17_P10996295 - -
Agilent Whole Chr array_18 19 21 A_17_P17156684 - -
Agilent Whole Chr array_18 19 21 A_17_P10996267 - -
Agilent Whole Chr array_18 19 21 A_17_P31635926 - -
Agilent Whole Chr array_18 19 21 A_17_P31635872 - -
Agilent Whole Chr array_18 19 21 A_17_P17156718 - -
Agilent Whole Chr array_18 19 21 A_17_P31635925 - -
Agilent Whole Chr array_18 19 21 A_17_P10996283 - -
Agilent Whole Chr array_18 19 21 A_17_P10996274 - -
Agilent Whole Chr array_18 19 21 A_17_P31635873 - -
Agilent Whole Chr array_18 19 21 A_17_P10996277 - -
Agilent Whole Chr array_18 19 21 A_17_P17156712 - -
Agilent Whole Chr array_18 19 21 A_17_P10996275 - -
Agilent Whole Chr array_18 19 21 A_17_P10996265 - -
Agilent Whole Chr array_18 19 21 A_17_P10996296 - -
Agilent Whole Chr array_18 19 21 A_17_P10996282 - -
Agilent Whole Chr array_18 19 21 A_17_P10996294 - -
Agilent Whole Chr array_18 19 21 A_17_P10996298 - -
Agilent Whole Chr array_18 19 21 A_17_P31635922 - -
Agilent Whole Chr array_18 19 21 A_17_P31635932 - -
Agilent Whole Chr array_18 19 21 A_17_P31635888 - -
Agilent Whole Chr array_18 19 21 A_17_P31635875 - -
Agilent Whole Chr array_18 19 21 A_17_P10996286 - -
Agilent Whole Chr array_18 19 21 A_17_P10996281 - -
Agilent Whole Chr array_18 19 21 A_17_P10996300 - -
Agilent Whole Chr array_18 19 21 A_17_P31635893 - -
Agilent Whole Chr array_18 19 21 A_17_P17156734 - -
Agilent Whole Chr array_18 19 21 A_17_P17156706 - -
Agilent Whole Chr array_18 19 21 A_17_P10996291 - -
Agilent Whole Chr array_18 19 21 A_17_P10996266 - -
Agilent Whole Chr array_18 19 21 A_17_P10996278 - -
Agilent Whole Chr array_18 19 21 A_17_P10996288 - -
Agilent Whole Chr array_18 19 21 A_17_P10996268 - -
Agilent Whole Chr array_18 19 21 A_17_P10996287 - -
Agilent Whole Chr array_18 19 21 A_17_P10996279 - -
Agilent Whole Chr array_18 19 21 A_17_P10996293 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31635905 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17156706 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10996281 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10996282 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10996280 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17156718 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17156717 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17156712 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31635875 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31635884 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10996266 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31635874 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10996278 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10996279 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31635872 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31635864 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10996277 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31635868 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10996275 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17156687 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10996272 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31635888 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10996274 - -
Agilent Whole Chr array_18 19 21 A_17_P31635922 - -
Agilent Whole Chr array_18 19 21 A_17_P10996273 - -
Agilent Whole Chr array_18 19 21 A_17_P31635874 - -
Agilent Whole Chr array_18 19 21 A_17_P10996264 - -
Agilent Whole Chr array_18 19 21 A_17_P10996265 - -
Agilent Whole Chr array_18 19 21 A_17_P31635905 - -
Agilent Whole Chr array_18 19 21 A_17_P10996282 - -
Agilent Whole Chr array_18 19 21 A_17_P17156687 - -
Agilent Whole Chr array_18 19 21 A_17_P10996299 - -
Agilent Whole Chr array_18 19 21 A_17_P31635864 - -
Agilent Whole Chr array_18 19 21 A_17_P10996276 - -
Agilent Whole Chr array_18 19 21 A_17_P31635884 - -
Agilent Whole Chr array_18 19 21 A_17_P31635873 - -
Agilent Whole Chr array_18 19 21 A_17_P10996280 - -
Agilent Whole Chr array_18 19 21 A_17_P10996268 - -
Agilent Whole Chr array_18 19 21 A_17_P31635893 - -
Agilent Whole Chr array_18 19 21 A_17_P10996267 - -
Agilent Whole Chr array_18 19 21 A_17_P10996295 - -
Agilent Whole Chr array_18 19 21 A_17_P10996274 - -
Agilent Whole Chr array_18 19 21 A_17_P10996297 - -
Agilent Whole Chr array_18 19 21 A_17_P10996290 - -
Agilent Whole Chr array_18 19 21 A_17_P10996278 - -
Agilent Whole Chr array_18 19 21 A_17_P10996277 - -
Agilent Whole Chr array_18 19 21 A_17_P31635872 - -
Agilent Whole Chr array_18 19 21 A_17_P31635926 - -
Agilent Whole Chr array_18 19 21 A_17_P10996275 - -
Agilent Whole Chr array_18 19 21 A_17_P31635861 - -
Agilent Whole Chr array_18 19 21 A_17_P10996300 - -
Agilent Whole Chr array_18 19 21 A_17_P10996292 - -
Agilent Whole Chr array_18 19 21 A_17_P10996291 - -
Agilent Whole Chr array_18 19 21 A_17_P31635910 - -
Agilent Whole Chr array_18 19 21 A_17_P10996279 - -
Agilent Whole Chr array_18 19 21 A_17_P10996281 - -
Agilent Whole Chr array_18 19 21 A_17_P17156712 - -
Agilent Whole Chr array_18 19 21 A_17_P10996283 - -
Agilent Whole Chr array_18 19 21 A_17_P17156706 - -
Agilent Whole Chr array_18 19 21 A_17_P17156734 - -
Agilent Whole Chr array_18 19 21 A_17_P10996266 - -
Agilent Whole Chr array_18 19 21 A_17_P31635875 - -
Agilent Whole Chr array_18 19 21 A_17_P31635868 - -
Agilent Whole Chr array_18 19 21 A_17_P17156684 - -
Agilent Whole Chr array_18 19 21 A_17_P10996294 - -
Agilent Whole Chr array_18 19 21 A_17_P10996284 - -
Agilent Whole Chr array_18 19 21 A_17_P10996289 - -
Agilent Whole Chr array_18 19 21 A_17_P10996269 - -
Agilent Whole Chr array_18 19 21 A_17_P31635932 - -
Agilent Whole Chr array_18 19 21 A_17_P31635890 - -
Agilent Whole Chr array_18 19 21 A_17_P10996288 - -
Agilent Whole Chr array_18 19 21 A_17_P17156717 - -
Agilent Whole Chr array_18 19 21 A_17_P10996298 - -
Agilent Whole Chr array_18 19 21 A_17_P10996287 - -
Agilent Whole Chr array_18 19 21 A_17_P31635918 - -
Agilent Whole Chr array_18 19 21 A_17_P10996296 - -
Agilent Whole Chr array_18 19 21 A_17_P10996285 - -
Agilent Whole Chr array_18 19 21 A_17_P31635862 - -
Agilent Whole Chr array_18 19 21 A_17_P31635925 - -
Agilent Whole Chr array_18 19 21 A_17_P10996272 - -
Agilent Whole Chr array_18 19 21 A_17_P10996293 - -
Agilent Whole Chr array_18 19 21 A_17_P10996301 - -
Agilent Whole Chr array_18 19 21 A_17_P10996286 - -
Agilent Whole Chr array_18 19 21 A_17_P31635907 - -
Agilent Whole Chr array_18 19 21 A_17_P17156718 - -
Agilent Whole Chr array_18 19 21 A_17_P31635888 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10996277 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17156687 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10996274 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31635868 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31635872 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10996281 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31635884 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17156706 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31635888 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10996272 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10996278 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31635905 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10996280 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10996279 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10996275 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17156712 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17156718 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17156717 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31635864 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10996282 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31635875 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31635874 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10996266 - -
Whole Human Genome Oligo Microarray G4112A A_23_P208238 - -
Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P208238 - -
Agilent Human 2 cDNA Microarray (G4101A) 1254325 - -
Agilent Human 2 cDNA Microarray (G4101A) 3390994 - -
Agilent Human 2 cDNA Microarray (G4101A) 1254325 - -
Agilent Human 2 cDNA Microarray (G4101A) 3390994 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P10996294 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P10996297 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P10996280 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P10996291 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P17156723 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P10996285 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P10996265 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P10996295 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P10996300 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P17156727 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P10996264 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P10996268 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P10996274 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P10996276 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P10996273 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P17156687 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P10996278 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P10996286 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P10996283 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P17156684 - -
Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P03463007 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.0 ST Array NR_023311_at - -
Affymetrix Human Gene 1.0 ST Array 8030959 - -
Affymetrix Human Gene 1.0 ST Array 8030958 - -
Affymetrix Human Gene 1.0 ST Array 8030960 - -
Affymetrix Human Gene 1.0 ST Array 8030961 - -
Affymetrix Human Gene 1.0 ST Array 8030957 - -
Affymetrix Human Gene 1.0 ST Array 8030962 - -
Affymetrix Human Gene 1.0 ST Array 8030963 - -
Affymetrix Human Gene 1.1 ST Array 8030954 - -
Affymetrix HT HG-U133+ PM Array Plate 207394_PM_at - -
Affymetrix Human Genome U219 Array 11752222_s_at - -
Affymetrix Human Genome U219 Array 11752222_PM_s_at - -
Affymetrix Human Clariom D Assay PSR1900202529.hg.1 NR_023311.1 Refseq
Affymetrix Human Clariom D Assay PSR1900202529.hg.1 TC1900011794.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900202529.hg.1 NR_023311.1 Refseq
Human Genome U133 Plus 2.0 Array 207394_PM_at - -
Human Genome U133 Plus 2.0 Array 207394_at - -
Human Genome U133A 2.0 Array 207394_at - -
Human Genome U133A 2.0 Array 207394_PM_at - -
AffymetrixPathwork Diagnostics Pathchip pw598937 - -
Human Genome U95 Set 34989_at - -
Human Genome U95 Set 34989_PM_at - -
Affymetrix Human Genome U133 Plus 2.0 Array 207394_at8 - -
Affymetrix Human Genome U133 Plus 2.0 Array 207394_at10 - -
Affymetrix Human Genome U133 Plus 2.0 Array 207394_at5 - -
Affymetrix Human Genome U133 Plus 2.0 Array 207394_at7 - -
Affymetrix Human Genome U133 Plus 2.0 Array 207394_at9 - -
Affymetrix Human Genome U133 Plus 2.0 Array 207394_at3 - -
Affymetrix Human Genome U133 Plus 2.0 Array 207394_at4 - -
Affymetrix Human Genome U133 Plus 2.0 Array 207394_at2 - -
Affymetrix Human Genome U133 Plus 2.0 Array 207394_at1 - -
Affymetrix Human Genome U133 Plus 2.0 Array 207394_at11 - -
Affymetrix Human Genome U133 Plus 2.0 Array 207394_at6 - -
Human Genome U133 Set 207394_at - -
Human Genome U133 Set 207394_PM_at - -
Array Name Probe IDs Targets Target Type
Arraystar Human LncRNA microarray V2.0 ASHG19A3A012969 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A012969 - -
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 195776 - -
Array Name Probe IDs Targets Target Type
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1723542 BM988340.1 human_transcriptome
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1723542 U09414.1 human_transcriptome
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1723542 NR_023311.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1723542 BM988340.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1723542 U09414.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1723542 NR_023311.1 human_transcriptome
Illumina human-6 v2.0 expression beadchip ILMN_1723542 BM988340.1 human_transcriptome
Illumina human-6 v2.0 expression beadchip ILMN_1723542 U09414.1 human_transcriptome
Illumina human-6 v2.0 expression beadchip ILMN_1723542 NR_023311.1 human_transcriptome
Illumina humanRef-8 v2.0 expression beadchip ILMN_1723542 BM988340.1 human_transcriptome
Illumina humanRef-8 v2.0 expression beadchip ILMN_1723542 U09414.1 human_transcriptome
Illumina humanRef-8 v2.0 expression beadchip ILMN_1723542 NR_023311.1 human_transcriptome
Illumina HumanRef-8 v3.0 expression beadchip ILMN_1723542 BM988340.1 human_transcriptome
Illumina HumanRef-8 v3.0 expression beadchip ILMN_1723542 U09414.1 human_transcriptome
Illumina HumanRef-8 v3.0 expression beadchip ILMN_1723542 NR_023311.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1723542 BM988340.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1723542 U09414.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1723542 NR_023311.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_1723542 BM988340.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_1723542 U09414.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_1723542 NR_023311.1 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_1723542 BM988340.1 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_1723542 U09414.1 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_1723542 NR_023311.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0