This product has been added to your favorites list. Go to
My Favorites
Please enter the information below and press OK to send your cart to Core Services for purchase.
Name
Phone number
Email
Budget number
PI Name
PI Email
Building and Room number
Pickup
Delivery
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs03466166_gH
See other FGFR2 GE Assays ›
Gene Symbol:
FGFR2
Gene Name:
fibroblast growth factor receptor 2
Gene Aliases:
BBDS, BEK, BFR-1, CD332, CEK3, CFD1, ECT1, JWS, K-SAM, KGFR, TK14, TK25
Chromosome Location:
Chr.10: 121478330 - 121598656 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs03466166_gH
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: FGFR2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_000141.4
NP_000132.3
7
1464
113
NM_001144913.1
NP_001138385.1
6
967
113
NM_001144915.1
NP_001138387.1
6
869
113
NM_001144916.1
NP_001138388.1
4
913
113
NM_001144917.1
NP_001138389.1
7
1464
113
NM_001144918.1
NP_001138390.1
5
1119
113
NM_001144919.1
NP_001138391.1
6
1197
113
NM_001320658.1
NP_001307587.1
7
1464
113
NM_022970.3
NP_075259.4
7
1464
113
NM_023029.2
NP_075418.1
5
700
113
NR_073009.1
5
1119
113
XM_006717708.2
XP_006717771.1
7
1662
113
XM_006717710.3
XP_006717773.1
7
1662
113
XM_006717711.2
XP_006717774.1
5
700
113
XM_006717712.2
XP_006717775.1
4
622
113
XM_017015920.1
XP_016871409.1
7
1662
113
XM_017015921.1
XP_016871410.1
7
1662
113
XM_017015922.1
XP_016871411.1
5
700
113
XM_017015923.1
XP_016871412.1
5
700
113
XM_017015924.1
XP_016871413.1
5
1317
113
XM_017015925.1
XP_016871414.1
5
1317
113
GenBank mRNA
AB030073.1
-
7
1235
113
AB030074.1
-
7
1235
113
AB030075.1
-
7
1235
113
AB030076.1
-
7
1235
113
AB030077.1
-
7
1235
113
AB030078.1
-
7
1235
113
AK294026.1
-
7
869
113
AK308621.1
-
4
913
113
BC039243.1
-
5
1119
5277896
113
CX873387.1
-
3
227
113
EF057068.1
-
5
550
113
EU826598.1
-
6
817
113
EU826599.1
-
6
817
113
EU826601.1
-
4
472
113
KP192323.1
-
5
634
113
KP192324.1
-
5
634
113
KP192326.1
-
5
634
113
KP192327.1
-
5
634
113
KP192328.1
-
6
901
113
KP192329.1
-
4
556
113
KP192330.1
-
5
634
113
KP192331.1
-
6
901
113
M55614.1
-
6
829
113
M80634.1
-
7
1235
113
M87770.1
-
7
1090
113
M87771.1
-
7
1304
113
M97193.1
-
7
1428
113
U11814.1
-
7
1075
113
X52832.1
-
7
996
113
X56191.1
-
6
841
113
Z71929.1
-
6
829
113
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length greater than or equal to 101
Protein Kinase
Mammalian Gene Collection
Detect Genomic DNA
Homo sapiens
Amplicon spans exons and probe does not span exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene FGFR2
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
MAPK cascade
angiogenesis
ureteric bud development
in utero embryonic development
epithelial to mesenchymal transition
positive regulation of mesenchymal cell proliferation
outflow tract septum morphogenesis
membranous septum morphogenesis
apoptotic process
cell-cell signaling
axonogenesis
positive regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
regulation of smoothened signaling pathway
post-embryonic development
embryonic pattern specification
organ morphogenesis
regulation of cell fate commitment
positive regulation of phospholipase activity
regulation of phosphatidylinositol 3-kinase signaling
morphogenesis of embryonic epithelium
peptidyl-tyrosine phosphorylation
orbitofrontal cortex development
ventricular zone neuroblast division
pyramidal neuron development
gland morphogenesis
positive regulation of Wnt signaling pathway
bone mineralization
lung development
epithelial cell differentiation
midbrain development
otic vesicle formation
hair follicle morphogenesis
lacrimal gland development
regulation of osteoblast proliferation
multicellular organism growth
organ growth
fibroblast growth factor receptor signaling pathway involved in negative regulation of apoptotic process in bone marrow
fibroblast growth factor receptor signaling pathway involved in hemopoiesis
fibroblast growth factor receptor signaling pathway involved in positive regulation of cell proliferation in bone marrow
fibroblast growth factor receptor signaling pathway involved in orbitofrontal cortex development
phosphatidylinositol-3-phosphate biosynthetic process
regulation of multicellular organism growth
regulation of fibroblast growth factor receptor signaling pathway
inner ear morphogenesis
odontogenesis
positive regulation of MAPK cascade
positive regulation of GTPase activity
cell fate commitment
regulation of osteoblast differentiation
positive regulation of cell cycle
positive regulation of transcription from RNA polymerase II promoter
protein autophosphorylation
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
lung alveolus development
mesodermal cell differentiation
embryonic digestive tract morphogenesis
embryonic organ morphogenesis
digestive tract development
embryonic organ development
reproductive structure development
positive regulation of smooth muscle cell proliferation
embryonic cranial skeleton morphogenesis
skeletal system morphogenesis
epidermis morphogenesis
branching morphogenesis of a nerve
mesenchymal cell differentiation
positive regulation of epithelial cell proliferation
negative regulation of epithelial cell proliferation
regulation of smooth muscle cell differentiation
positive regulation of cell division
ventricular cardiac muscle tissue morphogenesis
positive regulation of cardiac muscle cell proliferation
limb bud formation
bone development
bone morphogenesis
branching involved in prostate gland morphogenesis
branching involved in salivary gland morphogenesis
bud elongation involved in lung branching
lung lobe morphogenesis
lung-associated mesenchyme development
positive regulation of epithelial cell proliferation involved in lung morphogenesis
prostate gland morphogenesis
prostate epithelial cord elongation
prostate epithelial cord arborization involved in prostate glandular acinus morphogenesis
squamous basal epithelial stem cell differentiation involved in prostate gland acinus development
fibroblast growth factor receptor signaling pathway involved in mammary gland specification
lateral sprouting from an epithelium
mammary gland bud formation
epithelial cell proliferation involved in salivary gland morphogenesis
branch elongation involved in salivary gland morphogenesis
branching involved in labyrinthine layer morphogenesis
regulation of branching involved in prostate gland morphogenesis
regulation of morphogenesis of a branching structure
mesenchymal cell differentiation involved in lung development
mesenchymal cell proliferation involved in lung development
regulation of ERK1 and ERK2 cascade
positive regulation of ERK1 and ERK2 cascade
positive regulation of canonical Wnt signaling pathway
protein tyrosine kinase activity
fibroblast growth factor-activated receptor activity
Ras guanyl-nucleotide exchange factor activity
protein binding
ATP binding
heparin binding
1-phosphatidylinositol-3-kinase activity
fibroblast growth factor binding
protein homodimerization activity
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-4g6l8:80/100.66.76.118:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0