This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03645554_m1
See other CEACAM6 GE Assays ›
Gene Symbol:
CEACAM6
Gene Name:
carcinoembryonic antigen related cell adhesion molecule 6
Gene Aliases:
CD66c, CEAL, NCA
Chromosome Location:
Chr.19: 41755421 - 41772211 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03645554_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CEACAM6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002483.6 NP_002474.4 4 - 5 1074 67
GenBank mRNA AK312542.1 - 4 - 5 1059 67
BC005008.1 - 4 - 5 995 3640231 67
BT009774.1 - 4 - 5 955 67
M18216.1 - 4 - 5 1001 67
M18728.1 - 4 - 5 1005 67
M29541.1 - 4 - 5 1048 67

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CEACAM6

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
cell-cell signaling
leukocyte migration
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0