This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03654211_m1
See other TOM1 GE Assays ›
Gene Symbol:
TOM1
Gene Name:
target of myb1 membrane trafficking protein
Gene Aliases:
-
Chromosome Location:
Chr.22: 35299275 - 35347994 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03654211_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TOM1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_024195.1 2 - 3 262 104
GenBank mRNA AK295918.1 - 2 - 3 172 104
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DC336639.1 - 2 - 3 172 Chr. 22 - 35299894 - 35323874 [+] on Build GRCh38 104

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream HMG-box containing 4 HMGXB4 HMG2L1,
HMGBCG,
THC211630
-22.1kb Sense Chr. 22 - 35257274 - 35295807 on NCBI Build GRCh38
Downstream microRNA 3909 MIR3909 mir-3909 12.5kb Sense Chr. 22 - 35335640 - 35335758 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2200007196.hg.1
This assay targets the same location as HTA transcript cluster(s) TC22000257.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
endocytosis
protein transport
endosomal transport
protein binding
clathrin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR22004770.hg.1 ENST00000436462.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22004770.hg.1 ENST00000451197.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22004770.hg.1 NR_024194.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22004770.hg.1 NR_024195.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22004770.hg.1 TC22000257.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR2200150575.hg.1 TC2200007196.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2200150575.hg.1 ENST00000439512.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200150575.hg.1 ENST00000449508.1 Ensembl
Affymetrix Human Clariom D Assay PSR2200150575.hg.1 NR_024194.1 Refseq
Affymetrix Human Clariom D Assay PSR2200150575.hg.1 NR_024195.1 Refseq
Affymetrix Human Clariom D Assay PSR2200150575.hg.1 ENST00000439512.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200150575.hg.1 ENST00000449508.1 Ensembl
Affymetrix Human Clariom D Assay PSR2200150575.hg.1 NR_024194.1 Refseq
Affymetrix Human Clariom D Assay PSR2200150575.hg.1 NR_024195.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w8bxd:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0