This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03655075_s1
See other SLCO4A1 GE Assays ›
Gene Symbol:
SLCO4A1
Gene Name:
solute carrier organic anion transporter family member 4A1
Gene Aliases:
OATP-E, OATP1, OATP4A1, OATPE, OATPRP1, POAT, SLC21A12
Chromosome Location:
Chr.20: 62642445 - 62690700 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03655075_s1
Size
Availability Made To Order
Catalog # 4426961
Price 4,565.00
Your Price
Online offer:
4565.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLCO4A1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017027827.1 XP_016883316.1 12 3822 77
GenBank mRNA AK095985.1 - 1 1647 75

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream SLCO4A1 antisense RNA 1 SLCO4A1-AS1 -3.3kb Anti-sense Chr. 20 - 62663027 - 62666621 on NCBI Build GRCh38
Downstream neurotensin receptor 1 (high affinity) NTSR1 NTR 36.7kb Sense Chr. 20 - 62708770 - 62762771 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

thyroid hormone metabolic process
sodium-independent organic anion transport
thyroid hormone transport
sodium-independent organic anion transmembrane transporter activity
thyroid hormone transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline