This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03656683_s1
See other MEG3 GE Assays ›
Gene Symbol:
MEG3
Gene Name:
maternally expressed 3 (non-protein coding)
Gene Aliases:
FP504, GTL2, LINC00023, NCRNA00023, PRO0518, PRO2160, onco-lncRNA-83, prebp1
Chromosome Location:
Chr.14: 100826108 - 100861026 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03656683_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MEG3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_046473.1 7 9045 72
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BU929717.1 - 1 150 Chr. 14 - 100853512 - 100854285 [+] on Build GRCh38 72
GenBankmRNA BE045610.1 - 1 157 Chr. 14 - 100853505 - 100854012 [+] on Build GRCh38 72
GenBankmRNA BF956860.1 - 330 Chr. - - [-] on Build 72
GenBankmRNA AW341284.1 - 1 157 Chr. 14 - 100853505 - 100853976 [+] on Build GRCh38 72
GenBankmRNA BE327686.1 - 1 157 Chr. 14 - 100853505 - 100853974 [+] on Build GRCh38 72
GenBankmRNA AW445195.1 - 1 175 Chr. 14 - 100853492 - 100854011 [+] on Build GRCh38 72
GenBankmRNA AI694752.1 - 1 157 Chr. 14 - 100853505 - 100854094 [+] on Build GRCh38 72
GenBankmRNA AW341292.1 - 1 157 Chr. 14 - 100853505 - 100853981 [+] on Build GRCh38 72
GenBankmRNA BF108643.1 - 1 156 Chr. 14 - 100853506 - 100853964 [+] on Build GRCh38 72
GenBankmRNA AW615281.1 - 1 157 Chr. 14 - 100853505 - 100854010 [+] on Build GRCh38 72

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 770 MIR770 MIRN770,
hsa-mir-770
-1.0kb Sense Chr. 14 - 100852390 - 100852487 on NCBI Build GRCh38
Downstream microRNA 493 MIR493 MIRN493,
hsa-mir-493,
mir-493
15.1kb Sense Chr. 14 - 100869060 - 100869148 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1400008229.hg.1

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR1400158668.hg.1 NR_046473.1 Refseq
Affymetrix Human Clariom D Assay PSR1400158668.hg.1 NR_046473.1 Refseq
Affymetrix Human Clariom D Assay PSR1400158668.hg.1 TC1400008229.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1400158669.hg.1 NR_046473.1 Refseq
Affymetrix Human Clariom D Assay PSR1400158669.hg.1 NR_046473.1 Refseq
Affymetrix Human Clariom D Assay PSR1400158669.hg.1 TC1400008229.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 232265 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline