This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03657194_s1
See other LOC441086 GE Assays ›
Gene Symbol:
LOC441086 FAM169A
Gene Name:
uncharacterized LOC441086
family with sequence similarity 169 member A
Gene Aliases:
-
SLAP75
Chromosome Location:
Chr.5: 74865893 - 74867275 on Build GRCh38
Chromosome Location:
Chr.5: 74777574 - 74867994 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
-
Assay ID Hs03657194_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC441086
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_134288.1 1 1251 102
GenBank mRNA BC039003.1 - 1 1251 102
Gene Symbol: FAM169A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017009332.1 XP_016864821.1 1 102
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA HY220074.1 - 1 Chr. 5 - 74866844 - 74867267 [-] on Build GRCh38 102

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream NSA2,
ribosome biogenesis homolog
NSA2 CDK105,
HCL-G1,
HCLG1,
HUSSY-29,
HUSSY29,
TINP1
-89.0kb Sense Chr. 5 - 74765429 - 74776912 on NCBI Build GRCh38
Downstream glucosaminyl (N-acetyl) transferase 4,
core 2
GCNT4 C2GNT3 160.2kb Anti-sense Chr. 5 - 75027464 - 75030899 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017009332.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Affymetrix HT HG-U133+ PM Array Plate 1562920_PM_at - -
Human Genome U133 Plus 2.0 Array 1562920_PM_at - -
Human Genome U133 Plus 2.0 Array 1562920_at - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562920_at2 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562920_at3 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562920_at6 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562920_at1 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562920_at9 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562920_at10 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562920_at11 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562920_at5 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562920_at4 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562920_at7 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562920_at8 - -
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_1809404 - -
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1809404 - -
Illumina human-6 v2.0 expression beadchip ILMN_1809404 - -
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1809404 - -
Illumina HumanHT-12 V3.0 expression beadchip ILMN_1809404 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-fn2v6:80/100.66.76.37:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0